Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01703447_s1
          See other VMN1R180 GE Assays ›
          Gene Symbol:
          Vmn1r180
          Gene Name:
          vomeronasal 1 receptor 180
          Gene Aliases:
          Gm475, V1rd16
          Chromosome Location:
          Chr.7: 23952387 - 23953356 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01703447_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn1r180
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_206869.2 NP_996752.2 1 881 118
          GenBank mRNA BC150717.1 - 1 881 9087628 118

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn1r180

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P114317 BC150717.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P114317 NM_206869.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P114756 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P114317 BC150717.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P114317 NM_206869.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25003047 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P114756 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114317 BC150717.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114317 NM_206869.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P114756 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06656669 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06656655 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06656672 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06656677 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06656662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25003036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06656683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25003056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06656679 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10560781 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560784 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560745 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560731 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550825 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560796 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550784 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560729 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560800 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560798 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560743 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550761 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560790 - -
          Affymetrix Mouse Gene 1.1 ST Array 10550824 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5420551 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005420551 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1222469 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222469 - -
          mouse-6 v1.1 (Illumina) scl0232962.1_238-S - -
          Illumina Sentrix mouse-6.v1 bead array 5420551 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0232962.1_238-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5420551 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1222469 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1222469 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0