Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01703504_mH
          See other UGT3A1 GE Assays ›
          Gene Symbol:
          Ugt3a1
          Gene Name:
          UDP glycosyltransferases 3 family, polypeptide A1
          Gene Aliases:
          AI746432
          Chromosome Location:
          Chr.15: 9279826 - 9315038 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01703504_mH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ugt3a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207216.2 NP_997099.2 6 - 7 1423 92
          GenBank mRNA AK128903.1 - 6 - 7 1395 92
          AK143745.1 - 6 - 7 1401 92
          BC025940.1 - 6 - 7 1390 5100753 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500000072.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ugt3a1

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          flavonoid biosynthetic process
          cellular response to hormone stimulus
          flavonoid glucuronidation
          cellular response to genistein
          UDP-glycosyltransferase activity
          glucuronosyltransferase activity
          transferase activity
          transferase activity, transferring glycosyl groups
          transferase activity, transferring hexosyl groups

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1954216 AK128903.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1954216 AK143745.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1954216 BC025940.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1954216 NM_207216.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 581134 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 894117 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 563228 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 561152 AK011440.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 561152 AK147462.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 561152 AK147561.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 561152 AK159518.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 561152 AK217803.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 561152 BC030928.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 561152 BC047152.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 561152 BC095963.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 561152 NM_022012.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 590171 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 477220 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 621502 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 480714 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 169683 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 762222 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 727048 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 154191 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 911364 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 113033 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 100504 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 144042 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142757 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118650 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1954216 AK128903.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1954216 AK143745.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1954216 BC025940.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1954216 NM_207216.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30004976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30005008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30005002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30004978 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30004994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30004993 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30004992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30004985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30004982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30005009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30005000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30004980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30005003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30004998 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30005005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30004989 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30005012 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02970751 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02970759 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118650 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08280144 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142757 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08280145 - -
          Whole Mouse Genome G4122A A_51_P322396 BC025940.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118650 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142757 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12436847 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12436837 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12436831 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12436841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30004985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30004967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30004993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30004996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30004976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30004992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30004982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30004990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30004980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30004994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30004959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30004961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30004965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30004963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30004998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30004958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30004978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30005105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30005010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30005033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30005039 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12104288 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30005003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30004993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12436847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30005037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30005002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30004982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30004968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12005026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30005020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30004984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30004985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30005043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30004965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30005046 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30005026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30005040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30005048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30004978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30005028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30005045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12436841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12174233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12043764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30004975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30004980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30005008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30005035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12043758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30004963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30004962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30004992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30005000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30004996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30005024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30005014 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10423003 - -
          Affymetrix Mouse Gene 1.0 ST Array 10423014 - -
          Affymetrix Mouse Gene 1.0 ST Array 10427625 - -
          Affymetrix Mouse Gene 1.0 ST Array 10423004 - -
          Affymetrix Mouse Gene 1.0 ST Array 10423009 - -
          Affymetrix Mouse Gene 1.0 ST Array 10423007 - -
          Affymetrix Mouse Gene 1.0 ST Array 10427627 - -
          Affymetrix Mouse Gene 1.0 ST Array 10423006 - -
          Affymetrix Mouse Gene 1.0 ST Array 10423010 - -
          Affymetrix Mouse Gene 1.0 ST Array 10423008 - -
          Affymetrix Mouse Gene 1.0 ST Array 10427626 - -
          Affymetrix HT MG-430 PM Array Plate 1424493_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1451373_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10423002 - -
          Affymetrix Mouse Gene 1.1 ST Array 10423005 - -
          Affymetrix Mouse Gene 1.1 ST Array 10423011 - -
          Affymetrix Mouse Gene 1.1 ST Array 10427624 - -
          Mouse Genome 430 2.0 Array 1424493_s_at - -
          Mouse Genome 430 2.0 Array 1451373_PM_at - -
          Mouse Genome 430 2.0 Array 1424493_PM_s_at - -
          Mouse Genome 430 2.0 Array 1451373_at - -
          Affymetrix Mouse Clariom S Array PSR1500000652.mm.1 NM_207216.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000652.mm.1 NM_207216.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000652.mm.1 TC1500000072.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500000647.mm.1 NM_207216.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000647.mm.1 NM_207216.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000647.mm.1 TC1500000072.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500000648.mm.1 NM_207216.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000648.mm.1 NM_207216.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000648.mm.1 TC1500000072.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500000656.mm.1 NM_207216.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000656.mm.1 NM_207216.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000656.mm.1 TC1500000072.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500000654.mm.1 NM_207216.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000654.mm.1 NM_207216.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000654.mm.1 TC1500000072.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500000653.mm.1 NM_207216.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000653.mm.1 NM_207216.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000653.mm.1 TC1500000072.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500000644.mm.1 NM_207216.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000644.mm.1 NM_207216.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000644.mm.1 TC1500000072.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500000650.mm.1 NM_207216.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000650.mm.1 NM_207216.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000650.mm.1 TC1500000072.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500000651.mm.1 NM_207216.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000651.mm.1 NM_207216.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000651.mm.1 TC1500000072.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500000649.mm.1 NM_207216.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000649.mm.1 NM_207216.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000649.mm.1 TC1500000072.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500000655.mm.1 NM_207216.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000655.mm.1 NM_207216.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000655.mm.1 TC1500000072.mm.1 MTA TC
          Mouse Expression Set 430 1451373_at - -
          Mouse Expression Set 430 1424493_s_at - -
          Mouse Expression Set 430 1424493_PM_s_at - -
          Mouse Expression Set 430 1451373_PM_at - -
          Mouse Genome 430A 2.0 Array 1451373_at - -
          Mouse Genome 430A 2.0 Array 1424493_s_at - -
          Mouse Genome 430A 2.0 Array 1424493_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1451373_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424493_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1451373_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424493_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1451373_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 719772 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2918732 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2918732 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline