Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01707297_u1
          See other 4930433I11RIK GE Assays ›
          Gene Symbol:
          4930433I11Rik
          Gene Name:
          RIKEN cDNA 4930433I11 gene
          Gene Aliases:
          Gm481
          Chromosome Location:
          Chr.7: 40987609 - 40994833 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01707297_u1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930433I11Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207248.3 NP_997131.2 2 1285 148
          GenBank mRNA AK029662.1 - 2 1341 148
          BC150916.1 - 2 1285 9087827 148
          BC150921.1 - 2 1285 9087832 148

          Back To Top

          More Information


          Updated Mapping Information

          Based on assay re-evaluation in August 2010 with NCBI transcript and genome annotations (Genome NCBIM37.1), this assay is now annotated as a single-exon assay.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 4930433I11Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2060001 AK029662.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 891578 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 122233 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162988 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2060001 AK029662.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162988 - -
          Whole Mouse Genome G4122A A_52_P29862 AK029662.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P29862 BC150916.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P29862 BC150921.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P29862 NM_207248.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131034 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134476 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25075738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25075736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25075722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25075735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25075727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25075737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25075748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25075731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25075754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25075725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25075723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25075729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25075744 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25075735 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25075741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06627572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25075744 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25075758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25075736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25075746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06640556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06627575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25075768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25075761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25075737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06002635 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10552295 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552297 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552298 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562597 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562596 - -
          Affymetrix Mouse Gene 1.1 ST Array 10552294 - -
          Affymetrix Mouse Gene 1.1 ST Array 10562595 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6180647 - -
          Illumina mouse-6 v1.0 expression beadchip 6180647 - -
          Illumina mouse-6 v1.0 expression beadchip 6370097 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006180647 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006370097 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2687716 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2599632 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2599632 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2687716 - -
          mouse-6 v1.1 (Illumina) scl0243944.7_112-S - -
          mouse-6 v1.1 (Illumina) scl00243944.1_319-S - -
          Illumina Sentrix mouse-6.v1 bead array 6370097 - -
          Illumina Sentrix mouse-6.v1 bead array 6180647 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0243944.7_112-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00243944.1_319-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6180647 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6370097 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2599632 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2687716 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2599632 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2687716 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0