Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01716164_m1
          See other GM536 GE Assays ›
          Gene Symbol:
          Gm536
          Gene Name:
          predicted gene 536
          Gene Aliases:
          Gm1777
          Chromosome Location:
          Chr.16: 31617868 - 31638979 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01716164_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm536
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK029404.1 - 1 - 2 225 82

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 282189 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 131315 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 368265 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 367019 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 922809 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 454801 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 955292 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 108633 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P132193 AK029404.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08552516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648390 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648370 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13006516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648384 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648397 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648415 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648396 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648495 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648423 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648420 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648501 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12817486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648394 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648407 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12551208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648492 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12797338 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12817483 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12928233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12966306 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12603559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648375 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648491 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648496 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12522221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12803868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12869846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12793845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648385 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648416 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648387 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13006520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648505 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12817490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648371 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648483 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12510132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30648403 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 622078 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2537746 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline