Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01717269_m1
          See other FAM26D GE Assays ›
          Gene Symbol:
          Fam26d
          Gene Name:
          family with sequence similarity 26, member D
          Gene Aliases:
          4732454E20Rik, AI586164, Gm643
          Chromosome Location:
          Chr.10: 34038784 - 34044313 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01717269_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam26d
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081165.1 NP_001074634.1 1 - 2 611 64
          GenBank mRNA AK028763.1 - 1 - 2 605 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000001992.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          ion transmembrane transport
          cation transmembrane transport
          molecular_function
          cation channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2403159 AK028763.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160380 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2403159 AK028763.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P119882 NM_001081165.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P119882 AK028763.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27054765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08819802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08882946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08759903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08905524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08743918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054808 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054795 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08819801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08767060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08523198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08391523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08650609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08519619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08519620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054735 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08468279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27054796 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10368636 - -
          Affymetrix Mouse Gene 1.0 ST Array 10368637 - -
          Affymetrix HT MG-430 PM Array Plate 1436873_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10368635 - -
          Mouse Genome 430 2.0 Array 1436873_at - -
          Mouse Genome 430 2.0 Array 1436873_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 270711_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 270711_at - -
          Affymetrix Mouse Clariom S Array PSR1000014193.mm.1 NM_001081165.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000014193.mm.1 NM_001081165.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000014193.mm.1 TC1000001992.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000014194.mm.1 NM_001081165.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000014194.mm.1 NM_001081165.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000014194.mm.1 TC1000001992.mm.1 MTA TC
          Mouse Expression Set 430 1436873_PM_at - -
          Mouse Expression Set 430 1436873_at - -
          Murine Genome U74v2 Set 168168_PM_at - -
          Murine Genome U74v2 Set 168168_at - -
          Murine Genome U74v2 Set 168169_f_at - -
          Murine Genome U74v2 Set 168169_PM_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1436873_at - -
          Affymetrix HT Mouse Genome 430B Array 1436873_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 753118 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2533757 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3017166 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3017166 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline