Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01718147_m1
          See other 2810001G20RIK GE Assays ›
          Gene Symbol:
          2810001G20Rik
          Gene Name:
          RIKEN cDNA 2810001G20 gene
          Gene Aliases:
          AW045514
          Chromosome Location:
          Chr.11: 64079484 - 64083259 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01718147_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2810001G20Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033780.1 1 - 2 289 65
          GenBank mRNA AK005116.1 - 1 - 2 256 65
          AK007927.1 - 1 - 2 291 65
          AK012631.1 - 1 - 2 282 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000829.mm.1
          This assay targets the same location as RTA transcript cluster(s) TC1000000829.rn

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P81630 NR_033780.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P81630 AK005116.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P81630 AK007927.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P81630 AK012631.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P81630 NR_033780.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P81630 AK005116.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P81630 AK007927.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P81630 AK012631.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P81630 NR_033780.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P81630 AK005116.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P81630 AK007927.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P81630 AK012631.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P81630 NR_033780.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P81630 AK005116.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P81630 AK007927.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P81630 AK012631.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101734 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1455170_PM_at - -
          Mouse Genome 430 2.0 Array 1455170_PM_at - -
          Mouse Genome 430 2.0 Array 1455170_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66456_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66456_at - -
          Affymetrix Mouse Clariom S Array PSR1100006640.mm.1 NR_033780.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006640.mm.1 NR_033780.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006640.mm.1 TC1100000829.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100006643.mm.1 NR_033780.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006643.mm.1 NR_033780.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006643.mm.1 TC1100000829.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100006641.mm.1 NR_033780.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006641.mm.1 NR_033780.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006641.mm.1 TC1100000829.mm.1 MTA TC
          Mouse Expression Set 430 1455170_at - -
          Mouse Expression Set 430 1455170_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455170_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455170_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 804654 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline