| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10572961 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1430930_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10572960 |
- |
- |
| Mouse Genome 430 2.0 Array |
1430930_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1430930_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0800006370.mm.1 |
NR_028125.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006370.mm.1 |
NR_028125.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006370.mm.1 |
TC0800000864.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800006368.mm.1 |
NR_028125.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006368.mm.1 |
NR_028125.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006368.mm.1 |
TC0800000864.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800006369.mm.1 |
NR_028125.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006369.mm.1 |
NR_028125.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800006369.mm.1 |
TC0800000864.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1430930_PM_at |
- |
- |
| Mouse Expression Set 430 |
1430930_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1430930_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1430930_PM_at |
- |
- |