Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01718718_m1
          See other DGKEOS GE Assays ›
          Gene Symbol:
          Dgkeos
          Gene Name:
          diacylglycerol kinase, epsilon, opposite strand
          Gene Aliases:
          A930013B10Rik
          Chromosome Location:
          Chr.11: 89060751 - 89067884 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01718718_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dgkeos
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_110336.1 3 - 4 223 111
          GenBank mRNA AK044435.1 - 188 111
          BY080538.1 - 3 - 4 224 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001388.mm.1
          This assay targets the same location as RTA transcript cluster(s) TC1000001375.rn

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P367147 AK039819.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P367147 AK039819.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P367147 AK039819.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P367147 AK039819.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P367147 AK039819.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P367147 AK039819.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1456919_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1440233_PM_at - -
          Mouse Genome 430 2.0 Array 1456919_at - -
          Mouse Genome 430 2.0 Array 1440233_at - -
          Mouse Genome 430 2.0 Array 1456919_PM_at - -
          Mouse Genome 430 2.0 Array 1440233_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 414094_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 414094_at - -
          Affymetrix Mouse Clariom S Array PSR1100013022.mm.1 NR_110336.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013022.mm.1 TC1100001388.mm.1 MTA TC
          Mouse Expression Set 430 1440233_at - -
          Mouse Expression Set 430 1456919_PM_at - -
          Mouse Expression Set 430 1456919_at - -
          Mouse Expression Set 430 1440233_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1456919_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440233_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1456919_at - -
          Affymetrix HT Mouse Genome 430B Array 1440233_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106660390 - -
          Illumina mouse-6 v1.0 expression beadchip 106660390 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1251395 - -
          Illumina Sentrix mouse-6.v1 bead array 106660390 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106660390 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1251395 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline