| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10591752 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10591751 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10591750 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1442448_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10591749 |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0900016039.mm.1 |
NR_027981.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016039.mm.1 |
NR_027981.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016039.mm.1 |
TC0900001902.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900016043.mm.1 |
NR_027981.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016043.mm.1 |
NR_027981.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016043.mm.1 |
TC0900001902.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900016041.mm.1 |
NR_027981.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016041.mm.1 |
NR_027981.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016041.mm.1 |
TC0900001902.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900016042.mm.1 |
NR_027981.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016042.mm.1 |
NR_027981.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900016042.mm.1 |
TC0900001902.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1442448_at |
- |
- |
| Mouse Expression Set 430 |
1442448_PM_at |
- |
- |
| Murine Genome U74v2 Set |
130331_PM_at |
- |
- |
| Murine Genome U74v2 Set |
130331_at |
- |
- |