Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01720636_m1
          See other 9130410C08RIK GE Assays ›
          Gene Symbol:
          9130410C08Rik
          Gene Name:
          RIKEN cDNA 9130410C08 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01720636_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9130410C08Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK018667.1 - 1 - 2 182 195

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2319533 AK018667.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 436810 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 485376 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 208922 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 357530 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 78484 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 751866 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 618313 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 416058 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 541812 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 212873 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 498193 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 296303 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 391054 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 137965 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 259155 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 698079 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 264998 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 139297 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 955953 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 534 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 361345 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 812949 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 211006 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 3449 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 38302 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 132570 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120457 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124354 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163800 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2319533 AK018667.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00964121 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120457 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00964158 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05365452 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163800 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00964140 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124354 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05365358 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108686 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120457 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163800 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124354 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P00964140 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23221952 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222108 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23221943 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03439689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03580472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23221926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23221944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03287108 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222131 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03987710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03987712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03421981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222091 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222090 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04024583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23221992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04042934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03580482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03884413 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23221959 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03640692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03376809 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03605284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03937639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03310024 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03618395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03750698 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03605283 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03732826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23221932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23221935 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222001 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03281735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03559466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23221934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23221925 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222024 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23221955 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03937640 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222074 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03310023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04004145 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03580477 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03988810 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03593250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03439687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03763622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23221979 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03750697 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03282491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04042945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222068 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222120 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03281723 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03306480 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03987711 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222058 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222133 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222073 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03287112 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222101 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23221983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03937647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03281732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222117 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04042936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03750701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222127 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03306482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03439678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222050 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03421988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03421982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04042931 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04042940 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03281724 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222008 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03421983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03310027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04042930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222099 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222017 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222039 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03580484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23221949 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222107 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23221976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23221931 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23221927 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03987709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23221960 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222066 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03439688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03281725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222061 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23221958 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23221953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03937644 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23221938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04030266 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23221977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03370216 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03308767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23222122 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432309_PM_at - -
          Mouse Genome 430 2.0 Array 1432309_at - -
          Mouse Genome 430 2.0 Array 1432309_PM_at - -
          Mouse Expression Set 430 1432309_at - -
          Mouse Expression Set 430 1432309_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432309_at - -
          Affymetrix HT Mouse Genome 430B Array 1432309_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 621571 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103120288 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline