Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01720848_g1
          See other 3110004A20RIK GE Assays ›
          Gene Symbol:
          3110004A20Rik
          Gene Name:
          RIKEN cDNA 3110004A20 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01720848_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 3110004A20Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK019415.1 - 1 - 2 142 87

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent ABI

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09553229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09424253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09452263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735814 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09343753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735808 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09424265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09357891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09367406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27735821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09357894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09452260 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 444804 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-z8rtm:80/100.66.72.105:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline