Search
Search

F730043M19Rik| Gene Symbol: F730043M19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK089514.1 | - | 2 - 3 | 372 | 129 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cadherin-related family member 3 | Cdhr3 |
1110049B09Rik, BB172694, Cdh28 |
-49.6kb | Sense | Chr. 12 - 33033796 - 33093037 on NCBI Build GRCm38 |
| Downstream | ataxin 7-like 1 | Atxn7l1 |
2810423G08Rik, Atxn7l4 |
455.0 | Anti-sense | Chr. 12 - 33147639 - 33376408 on NCBI Build GRCm38 |
Set Membership: |
|