Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01721623_s1
          See other GM10190 GE Assays ›
          Gene Symbol:
          Gm10190
          Gene Name:
          predicted gene 10190
          Gene Aliases:
          ENSMUSG00000066938
          Chromosome Location:
          Chr.17: 80371857 - 80373542 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01721623_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm10190
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_028385.1 1 1510 146
          GenBank mRNA AK034626.1 - 1 1511 146

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2020981 NR_028385.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2020981 AK034626.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2020981 NM_001145452.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148639 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P148639 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P148639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13197723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31382667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31382670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31382681 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31382668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13567747 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31382674 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31382716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31382672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13197728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13186637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31382669 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31382691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31382684 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13186634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31382688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31382666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31382662 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.1 ST Array 10453139 - -
          Affymetrix Mouse Clariom S Array PSR1700023652.mm.1 NR_028385.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700023652.mm.1 NR_028385.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700023652.mm.1 TC1700002585.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700023651.mm.1 NR_028385.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700023651.mm.1 NR_028385.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700023651.mm.1 TC1700002585.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 667279 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 103130731 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245583 - -
          Illumina Sentrix mouse-6.v1 bead array 103130731 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103130731 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1245583 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0