Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01721667_m1
          See other 4930558C23RIK GE Assays ›
          Gene Symbol:
          4930558C23Rik
          Gene Name:
          RIKEN cDNA 4930558C23 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 95386458 - 95402841 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01721667_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930558C23Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015490.1 1 - 2 134 138
          GenBank mRNA AK006075.1 - 1 - 2 79 138
          AK006566.1 - 1 - 2 101 138
          AK016169.1 - 1 - 2 125 138
          BC049745.1 - 1 - 2 134 138

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300002478.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P246992 AK006075.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P246992 AK006566.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P246992 NR_015490.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P246992 AK016169.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P246992 BC049745.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P246992 AK006075.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P246992 AK006566.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P246992 NR_015490.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P246992 AK016169.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P246992 BC049745.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 530850 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 476445 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 403418 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 268905 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 575990 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 22913 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 742292 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 132745 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 91997 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 93019 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141978 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141978 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04994992 - -
          Whole Mouse Genome G4122A A_52_P246992 AK006075.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P246992 AK006566.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P246992 NR_015490.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P246992 AK016169.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P246992 BC049745.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P246992 AK006075.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P246992 AK006566.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P246992 NR_015490.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P246992 AK016169.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P246992 BC049745.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105933 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22343773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22343787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02486614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22343776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22343788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22343792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22343796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22343802 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22343780 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02770484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22343782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22343754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22343793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22343805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22343778 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22343790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02362028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22343781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22343775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22343786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22343807 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22343774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22343803 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22343801 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22343784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02770482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22343753 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22343791 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02362026 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300019207.mm.1 NR_015490.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300019207.mm.1 NR_015490.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300019207.mm.1 TC0300002478.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300019206.mm.1 NR_015490.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300019206.mm.1 NR_015490.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300019206.mm.1 TC0300002478.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 573432 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline