Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01721936_s1
          See other GM10129 GE Assays ›
          Gene Symbol:
          Gm10129
          Gene Name:
          predicted gene 10129
          Gene Aliases:
          ENSMUSG00000063303
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01721936_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm10129
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK037084.1 - 1 1125 79

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003678 AK037084.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003678 NM_023842.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119980 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003678 AK037084.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003678 NM_023842.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119980 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123426 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10702745 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11122734 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10752171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11122737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10698333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958859 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958805 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10698332 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10698324 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10808605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28958861 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 395742 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 101170180 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1237118 - -
          Illumina Sentrix mouse-6.v1 bead array 101170180 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101170180 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1237118 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline