Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01722569_g1
          See other GZMD GE Assays ›
          Gene Symbol:
          Gzmd
          Gene Name:
          granzyme D
          Gene Aliases:
          CCP5, Ctla-5, Ctla5
          Chromosome Location:
          Chr.14: 56129556 - 56132603 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01722569_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gzmd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010372.2 NP_034502.2 2 - 3 281 86
          GenBank mRNA AK131671.1 - 2 - 3 282 86
          BC141122.1 - 2 - 3 236 9055734 86
          BC145433.1 - 236 9053991 86
          BC145434.1 - 2 - 3 236 86
          J03255.1 - 2 - 3 296 86

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protease 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gzmd

          Panther Classification:

          Molecular Function -

          protease serine protease protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          immune response
          granzyme-mediated apoptotic signaling pathway
          protein processing
          cytolysis
          serine-type endopeptidase activity
          peptidase activity
          serine-type peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554650 AK131671.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554650 BC141122.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554650 BC145433.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554650 BC145434.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554650 J03255.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554650 AK131671.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554650 BC141122.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554650 BC145433.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554650 BC145434.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554650 J03255.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 9755 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 935960 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 329208 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 313347 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 42072 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554650 AK131671.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554650 BC141122.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554650 BC145433.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554650 BC145434.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554650 J03255.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554650 AK131671.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554650 BC141122.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554650 BC145433.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554650 BC145434.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554650 J03255.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P159042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621562 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08120368 - -
          Whole Mouse Genome G4122A A_52_P554650 AK131671.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554650 BC141122.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554650 BC145433.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554650 BC145434.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554650 J03255.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554650 AK131671.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554650 BC141122.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554650 BC145433.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554650 BC145434.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554650 J03255.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P159042 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124496 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11544113 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11658387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11255322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11526771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11526770 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621580 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10420279 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420275 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420277 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420276 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420269 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420278 - -
          Affymetrix HT MG-430 PM Array Plate 1420343_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1420344_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10420274 - -
          Affymetrix Mouse Gene 1.1 ST Array 10420268 - -
          Mouse Genome 430 2.0 Array 1420343_PM_at - -
          Mouse Genome 430 2.0 Array 1420343_at - -
          Mouse Genome 430 2.0 Array 1420344_x_at - -
          Mouse Genome 430 2.0 Array 1420344_PM_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14941_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14941_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1400016303.mm.1 NM_010372.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016303.mm.1 NM_010372.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016303.mm.1 TC1400002116.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016304.mm.1 NM_010372.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016304.mm.1 NM_010372.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016304.mm.1 TC1400002116.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016294.mm.1 NM_010373.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016294.mm.1 NM_010373.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016294.mm.1 TC1400002116.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016300.mm.1 NM_010372.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016300.mm.1 NM_010372.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016300.mm.1 TC1400002116.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016307.mm.1 NM_010372.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016307.mm.1 NM_010372.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016307.mm.1 TC1400002116.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016293.mm.1 NM_010373.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016293.mm.1 NM_010373.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016293.mm.1 TC1400002116.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016301.mm.1 NM_010372.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016301.mm.1 NM_010372.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016301.mm.1 TC1400002116.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016302.mm.1 NM_010372.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016302.mm.1 NM_010372.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016302.mm.1 TC1400002116.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016306.mm.1 NM_010372.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016306.mm.1 NM_010372.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016306.mm.1 TC1400002116.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016299.mm.1 NM_010372.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016299.mm.1 NM_010372.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016299.mm.1 TC1400002116.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016305.mm.1 NM_010372.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016305.mm.1 NM_010372.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016305.mm.1 TC1400002116.mm.1 MTA TC
          Mouse Expression Set 430 1420344_PM_x_at - -
          Mouse Expression Set 430 1420343_PM_at - -
          Mouse Expression Set 430 1420343_at - -
          Mouse Expression Set 430 1420344_x_at - -
          Murine Genome U74v2 Set 102688_PM_f_at - -
          Murine Genome U74v2 Set 102688_f_at - -
          Mouse Genome 430A 2.0 Array 1420343_PM_at - -
          Mouse Genome 430A 2.0 Array 1420344_x_at - -
          Mouse Genome 430A 2.0 Array 1420343_at - -
          Mouse Genome 430A 2.0 Array 1420344_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1420343_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420344_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1420343_at - -
          Affymetrix HT Mouse Genome 430A Array 1420344_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 847836 - -
          ABI Mouse Genome Survey Microarray 916732 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4920368 - -
          Illumina mouse-6 v1.0 expression beadchip 4920368 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004920368 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2678281 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2678281 - -
          mouse-6 v1.1 (Illumina) scl014941.4_46-S - -
          Illumina Sentrix mouse-6.v1 bead array 4920368 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl014941.4_46-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4920368 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2678281 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2987649 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2987649 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2678281 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline