Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01728657_g1
          See other GM13023 GE Assays ›
          Gene Symbol:
          Gm13023
          Gene Name:
          predicted gene 13023
          Gene Aliases:
          BC084594, MGC:91194, OTTMUSG00000010009
          Chromosome Location:
          Chr.4: 143789334 - 143795585 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01728657_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm13023
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007077.2 NP_001007078.2 2 - 3 406 125
          GenBank mRNA AK135972.1 - 2 - 3 389 125
          BC084594.1 - 2 - 3 387 30478700 125
          CA559385.1 - 2 - 3 376 125
          CF174133.1 - 2 - 3 414 125

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001837.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gm13023

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135865 NM_001007077.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135865 AK135972.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135865 BB285244.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135865 BC084594.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P175679 NM_001105254.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P175679 AK145346.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P175679 BC086484.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P175679 AK135749.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P175679 AK139641.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P175679 NM_001105254.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P175679 AK145346.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P175679 BC086484.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P175679 AK135749.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P175679 AK139641.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135865 NM_001007077.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135865 AK135972.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135865 BB285244.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135865 BC084594.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P175679 NM_001105254.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P175679 AK145346.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P175679 BC086484.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P175679 AK135749.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P175679 AK139641.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P175679 NM_001105254.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P175679 AK145346.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P175679 BC086484.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P175679 AK135749.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P175679 AK139641.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P175679 NM_001105254.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P175679 AK145346.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P175679 BC086484.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P175679 AK135749.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P175679 AK139641.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P175679 NM_001105254.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P175679 AK145346.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P175679 BC086484.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P175679 AK135749.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P175679 AK139641.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23376251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23376257 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01011161 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01011161 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03341963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23376204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23376202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23376292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03840522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03873025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23376214 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04011742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23376292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23376251 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10518190 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397550 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510089 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518189 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401826 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510085 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510088 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397548 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518188 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401825 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510087 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510086 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401827 - -
          Affymetrix Mouse Gene 1.0 ST Array 10397547 - -
          Affymetrix HT MG-430 PM Array Plate 1455813_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10518187 - -
          Affymetrix Mouse Gene 1.1 ST Array 10401824 - -
          Affymetrix Mouse Gene 1.1 ST Array 10510084 - -
          Affymetrix Mouse Gene 1.1 ST Array 10397545 - -
          Mouse Genome 430 2.0 Array 1455813_at - -
          Mouse Genome 430 2.0 Array 1455813_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 194227_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 194227_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400015115.mm.1 NM_001007077.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015115.mm.1 NM_001007077.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015115.mm.1 TC0400001837.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015119.mm.1 NM_001007077.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015119.mm.1 NM_001007077.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015119.mm.1 TC0400001837.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015121.mm.1 NM_001007077.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015121.mm.1 NM_001007077.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015121.mm.1 TC0400001837.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015118.mm.1 NM_001007077.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015118.mm.1 NM_001007077.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015118.mm.1 TC0400001837.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015117.mm.1 NM_001007077.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015117.mm.1 NM_001007077.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015117.mm.1 TC0400001837.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015120.mm.1 NM_001007077.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015120.mm.1 NM_001007077.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015120.mm.1 TC0400001837.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015114.mm.1 NM_001007077.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015114.mm.1 NM_001007077.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015114.mm.1 TC0400001837.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015116.mm.1 NM_001007077.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015116.mm.1 NM_001007077.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015116.mm.1 TC0400001837.mm.1 MTA TC
          Mouse Expression Set 430 1455813_at - -
          Mouse Expression Set 430 1455813_PM_at - -
          Mouse Genome 430A 2.0 Array 1455813_PM_at - -
          Mouse Genome 430A 2.0 Array 1455813_at - -
          Affymetrix HT Mouse Genome 430A Array 1455813_at - -
          Affymetrix HT Mouse Genome 430A Array 1455813_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1214493 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2900709 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2900709 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline