| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10419142 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10419143 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10419131 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10419112 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10419144 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10419130 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10419150 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10414267 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10419148 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10419135 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10419141 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10414266 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10419115 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10414268 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10419113 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10419133 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10419117 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10419134 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10419129 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10419114 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10419149 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10419145 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10414265 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10419116 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10419147 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10419140 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10419132 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10419111 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10419128 |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1400003855.mm.1 |
NM_198677.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003855.mm.1 |
NM_198677.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003855.mm.1 |
TC1400000466.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400003856.mm.1 |
NM_198677.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003856.mm.1 |
NM_198677.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003856.mm.1 |
TC1400000466.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400003858.mm.1 |
NM_198677.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003858.mm.1 |
NM_198677.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003858.mm.1 |
TC1400000466.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400003859.mm.1 |
NM_198677.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003859.mm.1 |
NM_198677.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003859.mm.1 |
TC1400000466.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400003857.mm.1 |
NM_198677.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003857.mm.1 |
NM_198677.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003857.mm.1 |
TC1400000466.mm.1 |
MTA TC |
| Murine Genome U74v2 Set |
170146_PM_r_at |
- |
- |
| Murine Genome U74v2 Set |
165888_PM_f_at |
- |
- |
| Murine Genome U74v2 Set |
170146_r_at |
- |
- |
| Murine Genome U74v2 Set |
165888_f_at |
- |
- |