Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01730228_gH
          See other ZFP758 GE Assays ›
          Gene Symbol:
          Zfp758
          Gene Name:
          zinc finger protein 758
          Gene Aliases:
          BC021442
          Chromosome Location:
          Chr.17: 22361453 - 22393909 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01730228_gH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp758
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145484.2 NP_663459.2 3 - 4 473 114
          GenBank mRNA AK028301.1 - 2 - 3 359 114
          BC022203.1 - 3 - 4 450 114
          BC037455.1 - 3 - 4 474 114

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp758

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1969273 BX515134.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1969273 AK028301.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1969273 NM_145484.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1969939 BC056632.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1969939 BC021442.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1969939 AK028301.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1969939 NM_145484.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 851566 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 768020 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 231425 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 603793 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 648586 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 581757 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 692228 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 638930 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 174008 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 92719 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 111436 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160539 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106923 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126305 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1969939 BC056632.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1969939 BC021442.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1969939 AK028301.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1969939 NM_145484.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1969273 BX515134.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1969273 AK028301.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1969273 NM_145484.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31089088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31089098 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31089092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31089090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31089087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31089105 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31089095 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31089089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31089091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31089104 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31089102 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31089101 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08735490 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08735502 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126305 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160539 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08735529 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106923 - -
          Whole Mouse Genome G4122A A_51_P494409 BX515134.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P494409 BC056632.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P494409 BC021442.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P494409 AK028301.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P494409 NM_145484.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P347770 BC022203.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P347770 BC037455.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P347770 BC056632.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P347770 BC021442.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P347770 AK028301.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106805 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160539 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106923 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08735490 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126305 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13463071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31089167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31089101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31089116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31089102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31089091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31089111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31089092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13375671 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31089113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31089089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13274836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31089098 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31089106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31089104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31089117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31089087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31089090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13073695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31089112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31089105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31089094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31089095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31089088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31089109 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13463070 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442267 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442266 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442269 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442264 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442265 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442268 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442263 - -
          Affymetrix HT MG-430 PM Array Plate 1451902_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10442262 - -
          Mouse Genome 430 2.0 Array 1451902_at - -
          Mouse Genome 430 2.0 Array 1451902_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 224598_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 224598_at - -
          Affymetrix Mouse Clariom S Array PSR1700002151.mm.1 NM_145484.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002151.mm.1 NM_145484.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002151.mm.1 TC1700000290.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002150.mm.1 NM_145484.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002150.mm.1 NM_145484.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002150.mm.1 TC1700000290.mm.1 MTA TC
          Mouse Expression Set 430 1451902_PM_at - -
          Mouse Expression Set 430 1451902_at - -
          Murine Genome U74v2 Set 104694_at - -
          Murine Genome U74v2 Set 104694_PM_at - -
          Mouse Genome 430A 2.0 Array 1451902_PM_at - -
          Mouse Genome 430A 2.0 Array 1451902_at - -
          Affymetrix HT Mouse Genome 430A Array 1451902_at - -
          Affymetrix HT Mouse Genome 430A Array 1451902_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2760059 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002760059 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2612506 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2612506 - -
          mouse-6 v1.1 (Illumina) scl00224598.1_163-S - -
          Illumina Sentrix mouse-6.v1 bead array 2760059 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00224598.1_163-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2760059 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2786125 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2786125 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2612506 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline