Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01731935_m1
          See other GGNBP1 GE Assays ›
          Gene Symbol:
          Ggnbp1
          Gene Name:
          gametogenetin binding protein 1
          Gene Aliases:
          0610031G08Rik, mdf
          Chromosome Location:
          Chr.17: 27018035 - 27036378 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01731935_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ggnbp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001251881.1 NP_001238810.1 6 - 7 831 65
          NM_027544.2 NP_081820.2 5 - 6 1260 65
          GenBank mRNA AB250379.1 - 5 - 6 1143 65
          AK077129.1 - 5 - 6 1143 65
          AY141192.1 - 5 - 6 1260 65
          BC049764.1 - 5 - 6 1121 6774330 65
          CO041745.1 - 1 - 2 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000429.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ggnbp1

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial fission
          multicellular organism development
          spermatogenesis
          cell differentiation
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1997465 BC049764.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1997465 AB250379.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1997465 AK077129.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1997465 AY141192.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1997465 NM_027544.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 214389 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 911706 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 151802 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 954362 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 171760 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108756 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1997465 BC049764.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1997465 AB250379.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1997465 AK077129.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1997465 AY141192.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1997465 NM_027544.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113823 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113833 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113827 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113837 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P108756 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P108756 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13178871 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13178874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13349690 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113859 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13178874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13061293 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13175144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13395991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13322907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13398915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13061294 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113817 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13266118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13395997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13061291 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13178880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13178873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13349691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113835 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10443126 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443127 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443130 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443128 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443124 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443125 - -
          Affymetrix HT MG-430 PM Array Plate 1433904_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1431199_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10443122 - -
          Mouse Genome 430 2.0 Array 1433904_PM_at - -
          Mouse Genome 430 2.0 Array 1433904_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70772_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70772_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700003726.mm.1 NM_001251881.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003726.mm.1 NM_001251881.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003726.mm.1 NM_027544.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003726.mm.1 NM_027544.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003726.mm.1 TC1700000429.mm.1 MTA TC
          Mouse Expression Set 430 1433904_at - -
          Mouse Expression Set 430 1433904_PM_at - -
          Murine Genome U74v2 Set 160644_at - -
          Murine Genome U74v2 Set 160644_PM_at - -
          Murine Genome U74v2 Set 169150_PM_i_at - -
          Murine Genome U74v2 Set 169150_i_at - -
          Mouse Genome 430A 2.0 Array 1433904_at - -
          Mouse Genome 430A 2.0 Array 1433904_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1433904_at - -
          Affymetrix HT Mouse Genome 430A Array 1433904_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 599654 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102100053 - -
          Illumina mouse-6 v1.0 expression beadchip 102100053 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2721198 - -
          mouse-6 v1.1 (Illumina) scl0070772.1_306-S - -
          Illumina Sentrix mouse-6.v1 bead array 102100053 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102100053 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2960522 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2721198 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2960522 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline