Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01732427_g1
          See other USMG5 GE Assays ›
          Gene Symbol:
          Usmg5
          Gene Name:
          upregulated during skeletal muscle growth 5
          Gene Aliases:
          2010301L15Rik
          Chromosome Location:
          Chr.19: 47083471 - 47090625 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01732427_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Usmg5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023211.4 NP_075700.2 3 - 4 234 70
          GenBank mRNA AK008345.1 - 3 - 4 235 70
          AK008511.1 - 1064 70
          AK038457.1 - 3 - 4 241 70
          AK088856.1 - 3 - 4 252 70
          BC024355.1 - 3 - 4 315 70
          BC087919.1 - 3 - 4 234 6545418 70

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Usmg5

          Panther Classification:

          Molecular Function -

          ATP synthase primary active transporter transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32147605 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32147604 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32147606 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P181365 AJ290947.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P181365 AJ290947.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463185 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 717246 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 195499 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 95117 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 570073 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 60672 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116927 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P181365 AJ290947.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P181365 AJ290947.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32147606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32147594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32147597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32147592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32147601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32147585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32147589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32147583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32147598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32147582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32147600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32147593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32147604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32147587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32147588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32147576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32147578 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32147595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32147607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32147603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32147575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32147591 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116927 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03608050 - -
          Whole Mouse Genome G4122A A_51_P181365 AJ290947.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P181365 AJ290947.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128922 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112002 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116927 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14403120 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14509856 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14212042 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32147604 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32147606 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32147605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32147599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32147592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32147606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32147576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32147578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14286807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32147580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32147603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14242057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32147589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14286806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32147590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32147585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32147571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32147598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14449913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32147599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32147565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32147593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32147595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32147573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14245933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14403120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32147591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32147594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32147564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14245931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32147582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14286805 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10468291 - -
          Affymetrix Mouse Gene 1.0 ST Array 10468290 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572814 - -
          Affymetrix Mouse Gene 1.0 ST Array 10468289 - -
          Affymetrix Mouse Gene 1.0 ST Array 10468288 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603834 - -
          Affymetrix HT MG-430 PM Array Plate 1448179_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10572813 - -
          Affymetrix Mouse Gene 1.1 ST Array 10603833 - -
          Affymetrix Mouse Gene 1.1 ST Array 10468287 - -
          Mouse Genome 430 2.0 Array 1448179_at - -
          Mouse Genome 430 2.0 Array 1448179_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900014368.mm.1 NM_023211.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014368.mm.1 NM_023211.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1900014368.mm.1 TC1900001621.mm.1 MTA TC
          Mouse Expression Set 430 1448179_at - -
          Mouse Expression Set 430 1448179_PM_at - -
          Murine Genome U74v2 Set 161452_f_at - -
          Murine Genome U74v2 Set 95718_PM_f_at - -
          Murine Genome U74v2 Set 161452_PM_f_at - -
          Murine Genome U74v2 Set 95718_f_at - -
          Murine Genome U74v2 Set 169791_PM_at - -
          Murine Genome U74v2 Set 169791_at - -
          Mouse Genome 430A 2.0 Array 1448179_PM_at - -
          Mouse Genome 430A 2.0 Array 1448179_at - -
          Affymetrix HT Mouse Genome 430A Array 1448179_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448179_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1580020 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001580020 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1219002 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2510420 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1219002 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2510420 - -
          mouse-6 v1.1 (Illumina) scl0066477.1_316-S - -
          Illumina Sentrix mouse-6.v1 bead array 1580020 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0066477.1_316-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1580020 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1219002 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1223794 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline