Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01733061_s1
          See other GM6981 GE Assays ›
          Gene Symbol:
          Gm6981
          Gene Name:
          glyceraldehyde-3-phosphate dehydrogenase pseudogene
          Gene Aliases:
          EG629557
          Chromosome Location:
          Chr.9: 52002061 - 52048705 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01733061_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm6981
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_023357.1 3 513 104
          GenBank mRNA AK132984.1 - 3 514 104

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961188 NR_023357.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961188 AK132984.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136832 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2077453 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2033530 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2124097 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 42217 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 827423 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 310395 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 901690 AK002795.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 901690 AK003106.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 901690 AK003205.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 901690 AK003558.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 901690 AK012604.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 901690 AK132329.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 901690 AK151614.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 901690 AK172322.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 901690 BC068180.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 901690 BC086774.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 901690 BC110308.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 901690 NM_026160.4 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 967798 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 184159 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 802605 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 718535 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 609889 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 30940 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 121884 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 899889 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 972721 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 380335 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 676119 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 299467 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 660710 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 194988 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 502161 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 284599 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 243842 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 854615 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 582903 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 565456 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 94317 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 689851 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 725963 AK038769.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 163421 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 127533 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 44823 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 21526 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 21209 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 169979 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2033530 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1961188 NR_023357.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1961188 AK132984.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136832 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2077453 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06788771 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01932650 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06788812 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06788771 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06788812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08143083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07569409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07871683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07500713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07776766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07600571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07515915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07964737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07550467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07871681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08190015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07550466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26512580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07954133 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10593491 - -
          Affymetrix Mouse Gene 1.0 ST Array 10447701 - -
          Affymetrix Mouse Gene 1.0 ST Array 10447700 - -
          Affymetrix Mouse Gene 1.1 ST Array 10593490 - -
          Affymetrix Mouse Gene 1.1 ST Array 10447699 - -
          Affymetrix Mouse Clariom S Array PSR0900019132.mm.1 NR_023357.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900019132.mm.1 TC0900002293.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900019127.mm.1 NR_023357.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900019127.mm.1 TC0900002293.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900019131.mm.1 NR_023357.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900019131.mm.1 TC0900002293.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900019129.mm.1 NR_023357.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900019129.mm.1 TC0900002293.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900019128.mm.1 NR_023357.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900019128.mm.1 TC0900002293.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline