Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01745212_s1
          See other GPANK1 GE Assays ›
          Gene Symbol:
          Gpank1
          Gene Name:
          G patch domain and ankyrin repeats 1
          Gene Aliases:
          Bat-4, Bat4, D17H6S54E, G5
          Chromosome Location:
          Chr.17: 35121473 - 35124815 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01745212_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpank1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001128597.1 NP_001122069.1 3 1291 120
          NM_032460.2 NP_115849.2 2 1089 120
          GenBank mRNA BC057902.1 - 3 1250 5027029 120
          BC120637.1 - 2 1090 40129560 120
          BC120639.1 - 2 1090 40129562 120

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpank1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          nucleic acid binding
          protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2022574 BC057902.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2022574 BY606764.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2022574 NM_001128597.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2022574 NM_032460.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 440928 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 252243 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 310609 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 122189 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106637 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2022574 BC057902.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2022574 BY606764.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2022574 NM_001128597.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2022574 NM_032460.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P136650 BC057902.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P136650 BC120637.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P136650 BC120639.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P136650 BY606764.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P136650 NM_001128597.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28854401 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28854402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157731 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07798491 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07798606 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07798516 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07798486 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106637 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07798594 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126819 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P122510 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02638712 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07798461 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07798566 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07798506 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07798644 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P103860 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07798582 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116102 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02638670 - -
          Whole Mouse Genome G4122A A_51_P136650 BC057902.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136650 BC120637.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136650 BC120639.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136650 BY606764.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136650 NM_001128597.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P103860 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07798461 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07798506 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116102 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02638719 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126819 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P122510 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106637 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13247230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157738 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13192768 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13192762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157731 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157727 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10403755 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444715 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444714 - -
          Affymetrix Mouse Gene 1.0 ST Array 10407925 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444716 - -
          Affymetrix HT MG-430 PM Array Plate 1423163_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1420284_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10403754 - -
          Affymetrix Mouse Gene 1.1 ST Array 10407924 - -
          Affymetrix Mouse Gene 1.1 ST Array 10444713 - -
          Mouse Genome 430 2.0 Array 1423163_at - -
          Mouse Genome 430 2.0 Array 1423163_PM_at - -
          Mouse Genome 430 2.0 Array 1420284_PM_at - -
          Mouse Genome 430 2.0 Array 1420284_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 81845_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 81845_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700006778.mm.1 NM_001128597.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006778.mm.1 NM_001128597.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006778.mm.1 NM_032460.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006778.mm.1 NM_032460.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006778.mm.1 TC1700000668.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700006779.mm.1 NM_001128597.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006779.mm.1 NM_001128597.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006779.mm.1 NM_032460.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006779.mm.1 NM_032460.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006779.mm.1 TC1700000668.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700006775.mm.1 NM_032460.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006775.mm.1 NM_032460.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006775.mm.1 TC1700000668.mm.1 MTA TC
          Mouse Expression Set 430 1420284_PM_at - -
          Mouse Expression Set 430 1423163_at - -
          Mouse Expression Set 430 1420284_at - -
          Mouse Expression Set 430 1423163_PM_at - -
          Murine Genome U74v2 Set 101416_f_at - -
          Murine Genome U74v2 Set 101415_PM_i_at - -
          Murine Genome U74v2 Set 101415_i_at - -
          Murine Genome U74v2 Set 161235_PM_f_at - -
          Murine Genome U74v2 Set 161235_f_at - -
          Murine Genome U74v2 Set 101416_PM_f_at - -
          Murine Genome U74v2 Set 170698_f_at - -
          Murine Genome U74v2 Set 170698_PM_f_at - -
          Murine Genome U74v2 Set 170233_at - -
          Murine Genome U74v2 Set 170233_PM_at - -
          Mouse Genome 430A 2.0 Array 1423163_at - -
          Mouse Genome 430A 2.0 Array 1420284_at - -
          Mouse Genome 430A 2.0 Array 1423163_PM_at - -
          Mouse Genome 430A 2.0 Array 1420284_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420284_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423163_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420284_at - -
          Affymetrix HT Mouse Genome 430A Array 1423163_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104060026 - -
          MouseWG-6 v2 Expression BeadChip 2690128 - -
          Illumina mouse-6 v1.0 expression beadchip 104060026 - -
          Illumina mouse-6 v1.0 expression beadchip 102370341 - -
          Illumina mouse-6 v1.0 expression beadchip 2690128 - -
          Illumina mouse-6 v1.0 expression beadchip 6220451 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002690128 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006220451 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2749976 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2621949 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2621949 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2749976 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1221493 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2466938 - -
          mouse-6 v1.1 (Illumina) scl0081845.1_1-S - -
          mouse-6 v1.1 (Illumina) scl49995.2.1_135-S - -
          mouse-6 v1.1 (Illumina) scl0081845.2_261-S - -
          mouse-6 v1.1 (Illumina) scl50790.1_36-S - -
          Illumina Sentrix mouse-6.v1 bead array 104060026 - -
          Illumina Sentrix mouse-6.v1 bead array 6220451 - -
          Illumina Sentrix mouse-6.v1 bead array 102370341 - -
          Illumina Sentrix mouse-6.v1 bead array 2690128 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0081845.1_1-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0081845.2_261-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102370341 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2690128 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6220451 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104060026 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2996046 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2621949 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2749976 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2749976 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2996046 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2621949 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline