Search
Search

E130006D01Rik| Gene Symbol: E130006D01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045832.1 | 5 | 568 | 119 | ||
| GenBank mRNA | AK087393.1 | - | 5 | 544 | 119 | |
| BB537478.2 | - | 3 | 296 | 119 | ||
Set Membership: |
|