Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01961800_s1
          See other OLFR1024 GE Assays ›
          Gene Symbol:
          Olfr1024
          Gene Name:
          olfactory receptor 1024
          Gene Aliases:
          MOR197-1
          Chromosome Location:
          Chr.2: 85904069 - 85905052 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01961800_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1024
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001005230.2 NP_001005230.2 1 533 121
          GenBank mRNA BC119447.1 - 1 533 40044257 121

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1024

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979625 NM_001005230.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979625 BC119447.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979625 NM_001005230.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979625 BC119447.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21399900 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399900 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01181999 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399887 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399876 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02184432 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399906 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399880 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01181982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399886 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01181991 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399903 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399871 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399895 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399866 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399869 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01181983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399897 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01905039 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399867 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21399859 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484554 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484553 - -
          Affymetrix Mouse Clariom S Array PSR0200033722.mm.1 NM_001005230.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033722.mm.1 NM_001005230.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033722.mm.1 TC0200003910.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160305 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160305 - -
          mouse-6 v1.1 (Illumina) GI_52421325-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3160305 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3160305 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline