Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01962366_s1
          See other ART2A-PS GE Assays ›
          Gene Symbol:
          Art2a-ps
          Gene Name:
          ADP-ribosyltransferase 2a, pseudogene
          Gene Aliases:
          ARTC2, Art2a, Ly92a, Rt-6, Rt6, Rt6-1
          Chromosome Location:
          Chr.7: 101552453 - 101560865 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01962366_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Art2a-ps
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007490.1 NP_031516.1 637 118
          NR_033804.1 3 707 118
          GenBank mRNA AF016462.1 - 637 118
          AF016464.1 - 2 637 118
          AK018555.1 - 3 708 118
          BC120753.1 - 731 40129676 118
          S79913.1 - 649 118
          X52991.1 - 637 118

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Art2a-ps

          Panther Classification:

          Molecular Function -

          transferase

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P306919 X52991.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P306919 NR_033804.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P306919 AF016462.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P306919 AF016464.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P306919 AK018555.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P306919 X52991.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P306919 NR_033804.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P306919 AF016462.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P306919 AF016464.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P306919 AK018555.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 275112 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 87554 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 186968 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 111568 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 156539 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 361912 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 725027 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 205666 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 2678 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 399409 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 528310 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 377857 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 563800 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 100325 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 26989 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 35665 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131162 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P306919 X52991.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P306919 NR_033804.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P306919 AF016462.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P306919 AF016464.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P306919 AK018555.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P306919 X52991.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P306919 NR_033804.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P306919 AF016462.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P306919 AF016464.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P306919 AK018555.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P306919 X52991.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P306919 NR_033804.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P306919 AF016462.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P306919 AF016464.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P306919 AK018555.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P306919 X52991.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P306919 NR_033804.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P306919 AF016462.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P306919 AF016464.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P306919 AK018555.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131162 - -
          Whole Mouse Genome G4122A A_51_P306919 X52991.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P306919 NR_033804.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P306919 AF016462.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P306919 AF016464.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P306919 AK018555.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P306919 X52991.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P306919 NR_033804.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P306919 AF016462.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P306919 AF016464.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P306919 AK018555.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124088 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25374262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06080859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06429947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06102618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06339432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05985095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06452596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06665655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25374254 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10565992 - -
          Affymetrix Mouse Gene 1.0 ST Array 10565995 - -
          Affymetrix Mouse Gene 1.0 ST Array 10565991 - -
          Affymetrix Mouse Gene 1.0 ST Array 10565993 - -
          Affymetrix HT MG-430 PM Array Plate 1423008_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10565990 - -
          Affymetrix Mouse Gene 1.1 ST Array 10565994 - -
          Mouse Genome 430 2.0 Array 1423008_PM_at - -
          Mouse Genome 430 2.0 Array 1423008_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11871_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11871_at - -
          Affymetrix Mouse Clariom S Array PSR0700032371.mm.1 TC0700003839.mm.1 MTA TC
          Mouse Expression Set 430 1423008_PM_at - -
          Mouse Expression Set 430 1423008_at - -
          Murine Genome U74v2 Set 99403_at - -
          Murine Genome U74v2 Set 99403_PM_at - -
          Mouse Genome 430A 2.0 Array 1423008_at - -
          Mouse Genome 430A 2.0 Array 1423008_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423008_at - -
          Affymetrix HT Mouse Genome 430A Array 1423008_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 454418 - -
          ABI Mouse Genome Survey Microarray 850060 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5050670 - -
          Illumina mouse-6 v1.0 expression beadchip 5050670 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005050670 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2680474 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2680474 - -
          mouse-6 v1.1 (Illumina) scl30965.3.1_50-S - -
          Illumina Sentrix mouse-6.v1 bead array 5050670 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl30965.3.1_50-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5050670 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2680474 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2680474 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline