Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01962535_s1
          See other GPR27 GE Assays ›
          Gene Symbol:
          Gpr27
          Gene Name:
          G protein-coupled receptor 27
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 99692679 - 99693818 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01962535_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpr27
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008158.1 NP_032184.1 1 74 148

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpr27

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          positive regulation of insulin secretion involved in cellular response to glucose stimulus
          positive regulation of phospholipase C-activating G-protein coupled receptor signaling pathway
          signal transducer activity
          G-protein coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24687705 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24687700 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24687698 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24687696 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24687697 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24687704 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24687703 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081040 NM_008158.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081040 NM_008158.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P260167 NM_008158.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P260167 NM_008158.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100877 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24687700 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24687696 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24687698 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24687705 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24687697 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24687704 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24687703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24687724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05012079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05528148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05227841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05528150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05528145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05040985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05579349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05530722 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05447399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05528141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05447402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05277225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05749429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05347656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05079184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05183799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05568880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687735 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05528147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05040986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05277226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24687761 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10540270 - -
          Affymetrix HT MG-430 PM Array Plate 1422154_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10540269 - -
          Mouse Genome 430 2.0 Array 1434848_at - -
          Mouse Genome 430 2.0 Array 1434848_PM_at - -
          Mouse Genome 430 2.0 Array 1422154_PM_at - -
          Mouse Genome 430 2.0 Array 1422154_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14761_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14761_at - -
          Affymetrix Mouse Clariom S Array PSR0600008888.mm.1 TC0600001146.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600008888.mm.1 NM_008158.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600008888.mm.1 NM_008158.1 Refseq
          Mouse Expression Set 430 1422154_at - -
          Mouse Expression Set 430 1422154_PM_at - -
          Murine Genome U74v2 Set 97151_at - -
          Murine Genome U74v2 Set 97151_PM_at - -
          Mouse Genome 430A 2.0 Array 1422154_PM_at - -
          Mouse Genome 430A 2.0 Array 1422154_at - -
          Affymetrix HT Mouse Genome 430A Array 1422154_at - -
          Affymetrix HT Mouse Genome 430A Array 1422154_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 736480 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3440037 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003440037 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2708377 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2708377 - -
          mouse-6 v1.1 (Illumina) scl1534.1.1_252-S - -
          Illumina Sentrix mouse-6.v1 bead array 3440037 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl1534.1.1_252-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3440037 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2708377 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3149700 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3070992 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2708377 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3070992 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3149700 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0