Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01964554_u1
          See other TREX2 GE Assays ›
          Gene Symbol:
          Trex2
          Gene Name:
          three prime repair exonuclease 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 73433705 - 73435343 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01964554_u1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trex2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011907.4 NP_036037.1 2 638 96
          GenBank mRNA AF151108.1 - 2 625 96
          BC104384.1 - 1 603 40050180 96
          BC104385.2 - 1 603 40050181 96

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trex2

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA catabolic process, exonucleolytic
          DNA metabolic process
          DNA repair
          cellular response to DNA damage stimulus
          magnesium ion binding
          nucleic acid binding
          nuclease activity
          exonuclease activity
          3'-5'-exodeoxyribonuclease activity
          exodeoxyribonuclease III activity
          hydrolase activity
          protein homodimerization activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2073094 NM_011907.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2073094 AF151108.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544929 BC047216.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544929 XM_488763.3 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544929 XM_489668.3 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 886073 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160254 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113215 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2073094 NM_011907.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2073094 AF151108.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P428341 AF151108.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32462611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32462592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32462593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32462606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32462618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32462590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32462595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32462589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32462599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32462588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32462610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32462607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32462605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32462591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32462614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32462615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32462617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32462612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32462609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32462603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32462598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32462587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32462616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32462602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32462619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32462597 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113215 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160254 - -
          Whole Mouse Genome G4122A A_51_P428341 AF151108.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137618 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160254 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113215 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14836141 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14836165 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14836149 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14836174 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14836142 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14836172 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14836139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32462586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14620228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32462590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32462583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32462606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15176854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32462582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32462614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14836167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32462598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32462592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32462595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32462605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15176857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32462603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15044340 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32462618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32462602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32462588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32462580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32462607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32462612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15176849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32462578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32462597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32462611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32462599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32462616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32462579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32462581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32462609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32462591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15044339 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32462587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32462610 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10605052 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605053 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605054 - -
          Affymetrix HT MG-430 PM Array Plate 1449367_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10605051 - -
          Mouse Genome 430 2.0 Array 1449367_at - -
          Mouse Genome 430 2.0 Array 1449367_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 24102_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 24102_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00014999.mm.1 NM_011907.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00014999.mm.1 NM_011907.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00014999.mm.1 TC0X00002427.mm.1 MTA TC
          Mouse Expression Set 430 1449367_at - -
          Mouse Expression Set 430 1449367_PM_at - -
          Murine Genome U74v2 Set 165582_PM_f_at - -
          Murine Genome U74v2 Set 171124_PM_at - -
          Murine Genome U74v2 Set 165582_f_at - -
          Murine Genome U74v2 Set 171124_at - -
          Mouse Genome 430A 2.0 Array 1449367_PM_at - -
          Mouse Genome 430A 2.0 Array 1449367_at - -
          Affymetrix HT Mouse Genome 430A Array 1449367_at - -
          Affymetrix HT Mouse Genome 430A Array 1449367_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 766350 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4920707 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004920707 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1225473 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1225473 - -
          mouse-6 v1.1 (Illumina) scl54164.2.1_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 4920707 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl54164.2.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4920707 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1225473 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1225473 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline