| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10405730 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10405729 |
- |
- |
| Mouse Genome 430 2.0 Array |
1447910_x_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1447910_PM_x_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1300004931.mm.1 |
NR_028430.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004931.mm.1 |
TC1300000746.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300004936.mm.1 |
NR_028430.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004936.mm.1 |
TC1300000746.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300004933.mm.1 |
NR_028430.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004933.mm.1 |
TC1300000746.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300004930.mm.1 |
NR_028430.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004930.mm.1 |
TC1300000746.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300004934.mm.1 |
NR_028430.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004934.mm.1 |
TC1300000746.mm.1 |
MTA TC |