Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01967666_s1
          See other GM13710 GE Assays ›
          Gene Symbol:
          Gm13710
          Gene Name:
          predicted gene 13710
          Gene Aliases:
          OTTMUSG00000013441
          Chromosome Location:
          Chr.2: 84500748 - 84506873 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01967666_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm13710
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046046.1 3 642 101

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2051199 AK044106.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 183427 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 605965 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 401572 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 38609 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 54478 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 421844 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 962036 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 193728 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 301603 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 547744 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 81463 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431833 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 812882 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 755919 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544914 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 733948 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 967091 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 957967 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 73057 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 401996 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 170329 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 24504 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 134963 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 147145 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 179653 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2051199 AK044106.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121941 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01545819 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393333 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393120 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01750594 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393118 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02032090 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393167 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393126 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393113 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01153918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393105 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01748082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393090 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393091 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393335 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393099 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393128 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02116061 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01753871 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393344 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02271114 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393116 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393119 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393114 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01153930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393342 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01481039 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01253342 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393107 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02271104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02271105 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393086 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393109 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01294287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02271116 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01750595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01193952 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01733769 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393111 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21393331 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200033406.mm.1 NR_046046.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033406.mm.1 NR_046046.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033406.mm.1 TC0200003867.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200033408.mm.1 NR_046046.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033408.mm.1 NR_046046.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033408.mm.1 TC0200003867.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200033405.mm.1 NR_046046.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033405.mm.1 NR_046046.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033405.mm.1 TC0200003867.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200033407.mm.1 NR_046046.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033407.mm.1 NR_046046.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033407.mm.1 TC0200003867.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 375256 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline