Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01968549_s1
          See other ARMCX3 GE Assays ›
          Gene Symbol:
          Armcx3
          Gene Name:
          armadillo repeat containing, X-linked 3
          Gene Aliases:
          1200004E24Rik, AI450003, ALEX3
          Chromosome Location:
          Chr.X: 134756568 - 134761457 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01968549_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Armcx3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027870.3 NP_082146.2 5 1380 167
          GenBank mRNA AK004598.1 - 5 1351 167
          AK030729.1 - 5 1353 167
          AK148876.1 - 1 842 168
          BC011101.1 - 1 887 3490748 167
          BC051113.1 - 5 1341 5249601 167
          BC108360.1 - 5 1320 167

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Armcx3

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular protein localization
          positive regulation of transcription from RNA polymerase II promoter
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006906 BC108360.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006906 AK148876.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006906 AK004598.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006906 AK030729.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006906 NM_027870.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2068977 AK148876.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2068977 AK219380.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2068977 BC011101.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2068977 BC051113.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2068977 AK004598.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 504502 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P164743 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2068977 AK148876.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2068977 AK219380.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2068977 BC011101.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2068977 BC051113.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2068977 AK004598.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006906 BC108360.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006906 AK148876.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006906 AK004598.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006906 AK030729.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006906 NM_027870.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32673529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32673526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32673533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32673524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32673536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32673527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32673525 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P164743 - -
          Whole Mouse Genome G4122A A_51_P404755 BC108360.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P404755 AK148876.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P404755 AK004598.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P404755 AK030729.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P404755 NM_027870.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P423380 AK148876.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P423380 BC011101.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P423380 BC051113.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P423380 AK004598.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P423380 AK030729.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101832 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119901 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P164743 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15004418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32673588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25508706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25508705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06233730 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673505 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15004426 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14639826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15143018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15004400 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15004415 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15004418 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673566 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10451854 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451852 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568365 - -
          Affymetrix Mouse Gene 1.0 ST Array 10601780 - -
          Affymetrix Mouse Gene 1.0 ST Array 10601779 - -
          Affymetrix Mouse Gene 1.0 ST Array 10601783 - -
          Affymetrix Mouse Gene 1.0 ST Array 10601781 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568364 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451853 - -
          Affymetrix Mouse Gene 1.0 ST Array 10601782 - -
          Affymetrix Mouse Gene 1.0 ST Array 10601784 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568366 - -
          Affymetrix Mouse Gene 1.0 ST Array 10601785 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451855 - -
          Affymetrix HT MG-430 PM Array Plate 1460359_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1424373_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10451851 - -
          Affymetrix Mouse Gene 1.1 ST Array 10601778 - -
          Affymetrix Mouse Gene 1.1 ST Array 10568363 - -
          Mouse Genome 430 2.0 Array 1424373_at - -
          Mouse Genome 430 2.0 Array 1460359_PM_at - -
          Mouse Genome 430 2.0 Array 1424373_PM_at - -
          Mouse Genome 430 2.0 Array 1460359_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71703_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71703_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00008072.mm.1 NM_027870.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008072.mm.1 NM_027870.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008072.mm.1 TC0X00001288.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00008073.mm.1 NM_027870.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008073.mm.1 NM_027870.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008073.mm.1 TC0X00001288.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036578.mm.1 TC0700004353.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036577.mm.1 TC0700004353.mm.1 MTA TC
          Mouse Expression Set 430 1460359_at - -
          Mouse Expression Set 430 1424373_PM_at - -
          Mouse Expression Set 430 1460359_PM_at - -
          Mouse Expression Set 430 1424373_at - -
          Mouse Genome 430A 2.0 Array 1460359_at - -
          Mouse Genome 430A 2.0 Array 1424373_PM_at - -
          Mouse Genome 430A 2.0 Array 1424373_at - -
          Mouse Genome 430A 2.0 Array 1460359_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 496368 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3290309 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003290309 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1234742 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1234742 - -
          mouse-6 v1.1 (Illumina) scl071703.5_40-S - -
          Illumina Sentrix mouse-6.v1 bead array 3290309 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl071703.5_40-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3290309 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1234742 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1234742 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl071703.5_40-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline