Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01971390_s1
          See other 2010204K13RIK GE Assays ›
          Gene Symbol:
          2010204K13Rik
          Gene Name:
          RIKEN cDNA 2010204K13 gene
          Gene Aliases:
          0610010M13Rik, Cip7
          Chromosome Location:
          Chr.X: 7411817 - 7422988 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01971390_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2010204K13Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027924.1 2 449 84
          NR_027925.1 2 459 84
          GenBank mRNA AF038507.1 - 2 413 84
          AK002502.1 - 798 84
          AK008437.1 - 2 450 84
          AK077356.1 - 2 493 84
          BC042200.1 - 2 412 84
          BC061056.1 - 2 460 84
          BC072610.1 - 2 373 84

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1987780 NR_027924.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1987780 AK077356.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P155805 NR_027924.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P155805 AF038507.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P155805 AK008437.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P155805 NR_027924.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P155805 AF038507.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P155805 AK008437.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444323 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 257471 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 319170 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 914511 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 822432 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 706675 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 765534 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 194848 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 290783 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 814478 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 844607 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 594437 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 16182 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 540171 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 528850 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 663693 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 630482 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 359255 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 642675 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 101983 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 774063 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 177766 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 45838 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119534 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161235 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P155805 NR_027924.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P155805 AF038507.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P155805 AK008437.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P155805 NR_027924.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P155805 AF038507.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P155805 AK008437.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1987780 NR_027924.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1987780 AK077356.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09225848 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119534 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161235 - -
          Whole Mouse Genome G4122A A_51_P179565 BC072610.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P179565 NR_027924.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P179565 NR_027925.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P179565 AF038507.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P179565 AK002502.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161235 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15206024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15132236 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238464 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238277 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238450 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238350 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238318 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15132231 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238285 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15193160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238493 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238437 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14870255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14767673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238323 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238309 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238291 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238465 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238319 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238467 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238338 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14850048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238337 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238501 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238466 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238345 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14767674 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14780131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238453 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238332 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238307 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238310 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238442 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14727812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238324 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238326 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238472 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238478 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14822337 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14850893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238308 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15132238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238356 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238286 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238503 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238474 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238328 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238505 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238445 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14996025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238257 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238473 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15196960 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238283 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238316 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14822335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15091159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238483 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15206027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238480 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14870264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238439 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15206026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238312 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238452 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14949186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238320 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238300 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238321 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15132233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238287 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238436 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238253 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14870260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15093371 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238455 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238496 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238476 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238454 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238301 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238280 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238451 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238458 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10603317 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603318 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603319 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603320 - -
          Affymetrix HT MG-430 PM Array Plate 1421498_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10603316 - -
          Mouse Genome 430 2.0 Array 1421498_PM_a_at - -
          Mouse Genome 430 2.0 Array 1421498_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68355_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68355_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00011165.mm.1 NR_027924.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011165.mm.1 NR_027924.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00011165.mm.1 TC0X00001773.mm.1 MTA TC
          Affymetrix HT Mouse Genome 430A Array 1421498_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1421498_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 932283 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ptcs5:80/100.66.79.248:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0