Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01971526_s1
          See other PET2 GE Assays ›
          Gene Symbol:
          Pet2
          Gene Name:
          plasmacytoma expressed transcript 2
          Gene Aliases:
          PC231, PC326, Pex3
          Chromosome Location:
          Chr.X: 89403848 - 89409689 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01971526_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pet2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008821.2 NP_032847.2 12 3786 161
          GenBank mRNA M95564.1 - 12 3791 161

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pet2

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960112 NM_008821.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960112 M95564.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 64 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 670286 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 547036 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 605286 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 111257 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171618 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960112 NM_008821.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960112 M95564.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32521214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32521224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32521217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32521227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32521216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32521198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32521223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32521220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32521212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32521213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32521218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32521226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32521210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32521225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32521221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32521206 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171618 - -
          Whole Mouse Genome G4122A A_52_P423238 NM_008821.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P423238 BC138080.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P423238 M95564.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108137 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125869 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32521240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521226 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521224 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521243 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15051360 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15107254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521237 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521248 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15060723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15051354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15060725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521235 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14944604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521231 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15051365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14862321 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14724794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521245 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14581094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521242 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521164 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14886618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14581099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32521178 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10605639 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605638 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605636 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605640 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605637 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605642 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605635 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605641 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605643 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605634 - -
          Affymetrix HT MG-430 PM Array Plate 1421566_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10605633 - -
          Mouse Genome 430 2.0 Array 1421566_PM_at - -
          Mouse Genome 430 2.0 Array 1421566_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18630_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18630_PM_at - -
          Affymetrix Murine Genome 6500A Array Msa10570_at - -
          Affymetrix Mouse Clariom S Array PSR0X00016315.mm.1 NM_008821.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016315.mm.1 NM_008821.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016315.mm.1 TC0X00002600.mm.1 MTA TC
          Mouse Expression Set 430 1421566_at - -
          Mouse Expression Set 430 1421566_PM_at - -
          Murine Genome U74v2 Set 96597_at - -
          Murine Genome U74v2 Set 96597_PM_at - -
          Mouse Genome 430A 2.0 Array 1421566_at - -
          Mouse Genome 430A 2.0 Array 1421566_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421566_at - -
          Affymetrix HT Mouse Genome 430A Array 1421566_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 430395 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5390100 - -
          Illumina mouse-6 v1.0 expression beadchip 5390100 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005390100 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2619442 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2619442 - -
          mouse-6 v1.1 (Illumina) scl018630.1_158-S - -
          Illumina Sentrix mouse-6.v1 bead array 5390100 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl018630.1_158-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5390100 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2821371 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2821371 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-zxk44:80/100.66.78.82:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0