Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01971861_g1
          See other RPS2 GE Assays ›
          Gene Symbol:
          Rps2
          Gene Name:
          ribosomal protein S2
          Gene Aliases:
          Llrep3, S4
          Chromosome Location:
          Chr.17: 24718121 - 24721927 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01971861_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rps2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008503.5 NP_032529.2 3 276 62
          GenBank mRNA AK004568.1 - 230 62
          AK012360.1 - 3 232 62
          AK019265.1 - 3 233 62
          AK151119.1 - 3 230 62
          AK151985.1 - 3 230 62
          AK152299.1 - 3 230 62
          AK159138.1 - 3 228 62
          AK163821.1 - 2 370 62
          AK166549.1 - 3 230 62
          AK166664.1 - 3 233 62
          AK166726.1 - 3 230 62
          AK167033.1 - 3 229 62
          AK167974.1 - 3 230 62
          AK168307.1 - 3 230 62
          AK168528.1 - 3 230 62
          AK168630.1 - 3 230 62
          AK169119.1 - 3 230 62
          BC087956.1 - 3 232 6749924 62
          BC091730.1 - 2 207 2651289 62
          BC094058.1 - 2 206 3675994 62
          BC106136.1 - 2 206 3676057 62
          BC110678.1 - 2 214 4011338 62

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK159138.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Amplicon spans exons and probe does not span exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rps2

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          ribosomal small subunit assembly
          formation of translation preinitiation complex
          translation
          positive regulation of transferase activity
          cellular response to interleukin-4
          RNA binding
          mRNA binding
          structural constituent of ribosome
          fibroblast growth factor binding
          enzyme binding
          protein complex binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31099200 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31099203 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2048454 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2070347 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 321844 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173934 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2070347 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2048454 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2104790 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P362278 BC092286.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P362278 BC093493.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P362278 BC094058.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P362278 BC094273.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P362278 BC106136.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P277592 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P182478 AK004568.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31099210 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173934 - -
          Whole Mouse Genome G4122A A_52_P182478 AK004568.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P277592 - -
          Whole Mouse Genome G4122A A_51_P362278 BC092286.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P362278 BC093493.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P362278 BC094058.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P362278 BC094273.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P362278 BC106136.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173934 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13136058 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13136057 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31099200 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31099203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31099179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384684 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13136056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14859052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384686 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099204 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15180655 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13136053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13136058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384681 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13258576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31099203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384690 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442581 - -
          Affymetrix Mouse Gene 1.0 ST Array 10459191 - -
          Affymetrix Mouse Gene 1.0 ST Array 10416188 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395923 - -
          Affymetrix Mouse Gene 1.0 ST Array 10459190 - -
          Affymetrix Mouse Gene 1.0 ST Array 10421684 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410888 - -
          Affymetrix Mouse Gene 1.0 ST Array 10599610 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606745 - -
          Affymetrix Mouse Gene 1.0 ST Array 10459189 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442583 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442582 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581307 - -
          Affymetrix Mouse Gene 1.0 ST Array 10599608 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488232 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505527 - -
          Affymetrix Mouse Gene 1.0 ST Array 10594321 - -
          Affymetrix Mouse Gene 1.0 ST Array 10599609 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534060 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438377 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402315 - -
          Affymetrix Mouse Gene 1.0 ST Array 10599611 - -
          Affymetrix Mouse Gene 1.0 ST Array 10516210 - -
          Affymetrix Mouse Gene 1.0 ST Array 10406279 - -
          Affymetrix Mouse Gene 1.0 ST Array 10459192 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541523 - -
          Affymetrix Mouse Gene 1.0 ST Array 10423362 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395924 - -
          Affymetrix HT MG-430 PM Array Plate 1431766_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10421683 - -
          Affymetrix Mouse Gene 1.1 ST Array 10594320 - -
          Affymetrix Mouse Gene 1.1 ST Array 10581306 - -
          Affymetrix Mouse Gene 1.1 ST Array 10402314 - -
          Affymetrix Mouse Gene 1.1 ST Array 10599607 - -
          Affymetrix Mouse Gene 1.1 ST Array 10410887 - -
          Affymetrix Mouse Gene 1.1 ST Array 10395922 - -
          Affymetrix Mouse Gene 1.1 ST Array 10416187 - -
          Affymetrix Mouse Gene 1.1 ST Array 10406278 - -
          Affymetrix Mouse Gene 1.1 ST Array 10423361 - -
          Affymetrix Mouse Gene 1.1 ST Array 10516209 - -
          Affymetrix Mouse Gene 1.1 ST Array 10459188 - -
          Affymetrix Mouse Gene 1.1 ST Array 10438376 - -
          Affymetrix Mouse Gene 1.1 ST Array 10534059 - -
          Affymetrix Mouse Gene 1.1 ST Array 10541522 - -
          Affymetrix Mouse Gene 1.1 ST Array 10505526 - -
          Affymetrix Mouse Gene 1.1 ST Array 10442580 - -
          Affymetrix Mouse Gene 1.1 ST Array 10488231 - -
          Affymetrix Mouse Gene 1.1 ST Array 10606744 - -
          Mouse Genome 430 2.0 Array 1422156_a_at - -
          Mouse Genome 430 2.0 Array 1431765_a_at - -
          Mouse Genome 430 2.0 Array 1431765_PM_a_at - -
          Mouse Genome 430 2.0 Array 1422156_PM_a_at - -
          Mouse Genome 430 2.0 Array 1431766_PM_x_at - -
          Mouse Genome 430 2.0 Array 1431766_x_at - -
          Affymetrix Mouse Clariom S Array PSR1700002703.mm.1 NM_008503.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002703.mm.1 NM_008503.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002703.mm.1 TC1700000349.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002710.mm.1 NM_008503.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002710.mm.1 NM_008503.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002710.mm.1 TC1700000349.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002718.mm.1 NM_008503.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002718.mm.1 NM_008503.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002718.mm.1 TC1700000349.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002717.mm.1 NM_008503.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002717.mm.1 NM_008503.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002717.mm.1 TC1700000349.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002709.mm.1 NM_008503.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002709.mm.1 NM_008503.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002709.mm.1 TC1700000349.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002714.mm.1 NM_008503.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002714.mm.1 NM_008503.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002714.mm.1 TC1700000349.mm.1 MTA TC
          Mouse Expression Set 430 1431765_a_at - -
          Mouse Expression Set 430 1431766_PM_x_at - -
          Mouse Expression Set 430 1422156_a_at - -
          Mouse Expression Set 430 1431765_PM_a_at - -
          Mouse Expression Set 430 1431766_x_at - -
          Mouse Expression Set 430 1422156_PM_a_at - -
          Murine Genome U74v2 Set 100686_PM_at - -
          Murine Genome U74v2 Set 100686_at - -
          Mouse Genome 430A 2.0 Array 1431766_x_at - -
          Mouse Genome 430A 2.0 Array 1431766_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1422156_a_at - -
          Mouse Genome 430A 2.0 Array 1431765_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1422156_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1431765_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1431766_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1431766_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 895573 - -
          ABI Mouse Genome Survey Microarray 896804 - -
          ABI Mouse Genome Survey Microarray 901987 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1340647 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001340647 - -
          mouse-6 v1.1 (Illumina) scl016898.7_20-S - -
          Illumina Sentrix mouse-6.v1 bead array 1340647 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl016898.7_20-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1340647 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2946616 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2717549 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2946616 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl016898.7_20-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline