Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01973494_s1
          See other PSORS1C2 GE Assays ›
          Gene Symbol:
          Psors1c2
          Gene Name:
          psoriasis susceptibility 1 candidate 2 (human)
          Gene Aliases:
          AI448740, Pcg, SPR1
          Chromosome Location:
          Chr.17: 35532207 - 35534647 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01973494_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psors1c2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020576.2 NP_065601.2 2 787 130
          GenBank mRNA AF159091.1 - 2 719 130
          AF484421.1 - 2 746 130
          AK137759.1 - 2 788 130

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psors1c2

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1988714 AK137759.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1988714 AF159091.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1988714 AF484421.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1988714 NM_020576.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165282 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106419 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P111639 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1988714 AK137759.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1988714 AF159091.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1988714 AF484421.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1988714 NM_020576.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P212958 AK137759.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P212958 BC110561.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P212958 AF159091.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P212958 AF484421.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P212958 NM_020576.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31159584 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165282 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P111639 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106419 - -
          Whole Mouse Genome G4122A A_51_P212958 AK137759.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P212958 BC110561.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P212958 AF159091.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P212958 AF484421.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P212958 NM_020576.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100662 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P111639 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106419 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165282 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13138427 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13327958 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13327963 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13327969 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13327951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31159594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13272572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13140436 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13543706 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13082203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13382571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13281506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13327955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13281512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31159618 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10444880 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444882 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444881 - -
          Affymetrix HT MG-430 PM Array Plate 1420467_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10444879 - -
          Mouse Genome 430 2.0 Array 1420467_PM_at - -
          Mouse Genome 430 2.0 Array 1420467_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57390_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57390_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700007128.mm.1 NM_020576.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007128.mm.1 NM_020576.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007128.mm.1 TC1700000693.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700007131.mm.1 NM_020576.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007131.mm.1 NM_020576.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007131.mm.1 TC1700000693.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700007130.mm.1 NM_020576.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007130.mm.1 NM_020576.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007130.mm.1 TC1700000693.mm.1 MTA TC
          Mouse Expression Set 430 1420467_PM_at - -
          Mouse Expression Set 430 1420467_at - -
          Mouse Genome 430A 2.0 Array 1420467_PM_at - -
          Mouse Genome 430A 2.0 Array 1420467_at - -
          Affymetrix HT Mouse Genome 430A Array 1420467_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420467_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 882433 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 770133 - -
          Illumina mouse-6 v1.0 expression beadchip 770133 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000770133 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1236058 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1236058 - -
          mouse-6 v1.1 (Illumina) scl50773.2.1_32-S - -
          Illumina Sentrix mouse-6.v1 bead array 770133 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50773.2.1_32-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 770133 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1236058 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1236058 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline