Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01973771_s1
          See other GM5483 GE Assays ›
          Gene Symbol:
          Gm5483
          Gene Name:
          predicted gene 5483
          Gene Aliases:
          EG433016
          Chromosome Location:
          Chr.16: 36184212 - 36188111 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01973771_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm5483
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001082547.1 NP_001076016.1 3 349 92

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          protease inhibitor

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401907 BB843120.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401907 BC147081.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401907 BC147082.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401907 AK089257.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401907 NM_001082547.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401907 BB843120.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401907 BC147081.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401907 BC147082.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401907 AK089257.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401907 NM_001082547.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401907 BB843120.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401907 BC147081.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401907 BC147082.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401907 AK089257.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401907 NM_001082547.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401907 BB843120.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401907 BC147081.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401907 BC147082.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401907 AK089257.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401907 NM_001082547.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08563922 - -
          Whole Mouse Genome G4122A A_51_P401907 BB843120.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401907 BC147081.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401907 BC147082.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401907 AK089257.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401907 NM_001082547.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401907 BB843120.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401907 BC147081.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401907 BC147082.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401907 AK089257.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401907 NM_001082547.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30674062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30674013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30674064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30674113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30674136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30674080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12504726 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30674101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30674081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30674025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30674031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30674037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30674080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30674045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30674019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30674051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12803799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30674036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30674017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12810865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12912657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30674094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30674064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12786090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30674062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12614205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12790625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30674035 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10608637 - -
          Affymetrix Mouse Gene 1.1 ST Array 10608637 - -
          Affymetrix Mouse Clariom S Array PSR1600004840.mm.1 NM_001082547.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600004840.mm.1 NM_001082547.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600004840.mm.1 TC1600000508.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600004843.mm.1 NM_001082547.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600004843.mm.1 NM_001082547.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600004843.mm.1 TC1600000508.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600004844.mm.1 NM_001082547.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600004844.mm.1 NM_001082547.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600004844.mm.1 TC1600000508.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600004841.mm.1 NM_001082547.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600004841.mm.1 NM_001082547.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600004841.mm.1 TC1600000508.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 774122 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2995688 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2995688 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0