Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01973813_g1
          See other A930005H10RIK GE Assays ›
          Gene Symbol:
          A930005H10Rik
          Gene Name:
          RIKEN cDNA A930005H10 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 115881578 - 115888130 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01973813_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A930005H10Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015487.1 1 334 59
          NR_027895.1 1 324 59
          GenBank mRNA AK020812.1 - 1 335 59
          AK081943.1 - 1 302 59
          AK137339.1 - 1 302 59

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P125862 AK137339.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P125862 NR_015487.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P125862 NR_027895.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P125862 AK020812.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P125862 AK081943.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 196011 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 427074 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 234797 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 189903 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 662393 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 8365 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158369 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158369 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125862 AK137339.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125862 NR_015487.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125862 NR_027895.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125862 AK020812.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125862 AK081943.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158369 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02973793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22446054 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02973796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22446049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22446051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22446048 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10501592 - -
          Affymetrix HT MG-430 PM Array Plate 1457709_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1441263_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1460135_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1441264_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10501591 - -
          Mouse Genome 430 2.0 Array 1460135_at - -
          Mouse Genome 430 2.0 Array 1441263_a_at - -
          Mouse Genome 430 2.0 Array 1457709_PM_a_at - -
          Mouse Genome 430 2.0 Array 1441263_PM_a_at - -
          Mouse Genome 430 2.0 Array 1441264_PM_x_at - -
          Mouse Genome 430 2.0 Array 1441264_x_at - -
          Mouse Genome 430 2.0 Array 1460135_PM_at - -
          Mouse Genome 430 2.0 Array 1457709_a_at - -
          Affymetrix Mouse Clariom S Array PSR0300022023.mm.1 NR_015487.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300022023.mm.1 NR_015487.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300022023.mm.1 NR_027894.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300022023.mm.1 NR_027894.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300022023.mm.1 NR_027895.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300022023.mm.1 NR_027895.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300022023.mm.1 TC0300002799.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300022021.mm.1 NR_027895.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300022021.mm.1 NR_027895.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300022021.mm.1 TC0300002799.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300022017.mm.1 NR_015487.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300022017.mm.1 NR_015487.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300022017.mm.1 NR_027894.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300022017.mm.1 NR_027894.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300022017.mm.1 TC0300002799.mm.1 MTA TC
          Mouse Expression Set 430 1460135_PM_at - -
          Mouse Expression Set 430 1460135_at - -
          Mouse Expression Set 430 1457709_a_at - -
          Mouse Expression Set 430 1441264_PM_x_at - -
          Mouse Expression Set 430 1441263_PM_a_at - -
          Mouse Expression Set 430 1441264_x_at - -
          Mouse Expression Set 430 1438823_PM_at - -
          Mouse Expression Set 430 1438823_at - -
          Mouse Expression Set 430 1457709_PM_a_at - -
          Mouse Expression Set 430 1441263_a_at - -
          Murine Genome U74v2 Set 165909_PM_at - -
          Murine Genome U74v2 Set 165909_at - -
          Affymetrix HT Mouse Genome 430B Array 1441263_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1460135_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1460135_at - -
          Affymetrix HT Mouse Genome 430B Array 1441264_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1457709_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1457709_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1441264_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1441263_a_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline