Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01973893_g1
          See other EEF1A1 GE Assays ›
          Gene Symbol:
          Eef1a1
          Gene Name:
          eukaryotic translation elongation factor 1 alpha 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 78478453 - 78481724 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01973893_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eef1a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010106.2 NP_034236.2 2 198 117
          GenBank mRNA AK003188.1 - 2 146 117
          AK011771.1 - 2 145 117
          AK012341.1 - 2 146 117
          AK032914.1 - 2 144 117
          AK050635.1 - 2 145 117
          AK076696.1 - 2 146 117
          AK081725.1 - 2 146 117
          AK083361.1 - 2 146 117
          AK088041.1 - 2 146 117
          AK133725.1 - 2 146 117
          AK135448.1 - 2 147 117
          AK137223.1 - 2 146 117
          AK141243.1 - 2 146 117
          AK150798.1 - 2 146 117
          AK151479.1 - 2 147 117
          AK151773.1 - 2 146 117
          AK152810.1 - 2 146 117
          AK152993.1 - 2 146 117
          AK153387.1 - 2 146 117
          AK153402.1 - 2 146 117
          AK153851.1 - 2 146 117
          AK162049.1 - 2 146 117
          AK162313.1 - 2 145 117
          AK162440.1 - 2 150 117
          AK163799.1 - 2 146 117
          AK164113.1 - 2 148 117
          AK164671.1 - 2 146 117
          AK165938.1 - 2 146 117
          AK166021.1 - 2 146 117
          AK166033.1 - 2 146 117
          AK166058.1 - 2 146 117
          AK166089.1 - 2 146 117
          AK166615.1 - 2 146 117
          AK166807.1 - 2 146 117
          AK167843.1 - 2 146 117
          AK168424.1 - 2 146 117
          AK168467.1 - 2 152 117
          BC003969.1 - 1 116 117
          BC004005.1 - 1 107 3492770 117
          BC004067.1 - 1 135 3590965 117
          BC005660.1 - 1 136 3588966 117
          BC018223.1 - 2 134 4012653 117
          BC018485.1 - 2 148 3491827 117
          BC050124.1 - 1691 117
          BC068258.1 - 1943 117
          BC083069.1 - 2 134 6310448 117
          BC092053.1 - 1 124 4008574 117
          BC092276.1 - 1 129 3490007 117
          BC092292.1 - 1 90 117
          BC095965.1 - 1 124 6310404 117
          BC108391.1 - 2 131 30242277 117
          BY227095.1 - 2 175 117
          M22432.1 - 2 137 117
          X13661.1 - 1 117 117

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Eef1a1

          Panther Classification:

          Molecular Function -

          translation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          translational elongation
          protein homooligomerization
          cellular response to epidermal growth factor stimulus
          tRNA binding
          nucleotide binding
          mRNA binding
          translation elongation factor activity
          GTPase activity
          protein binding
          GTP binding
          protein kinase binding
          poly(A) RNA binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900022513.mm.1 NM_010106.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900022513.mm.1 NM_010106.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900022513.mm.1 TC0900002727.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900022509.mm.1 NM_010106.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900022509.mm.1 NM_010106.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900022509.mm.1 TC0900002727.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900022520.mm.1 NM_010106.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900022520.mm.1 NM_010106.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900022520.mm.1 TC0900002727.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900022511.mm.1 NM_010106.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900022511.mm.1 NM_010106.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900022511.mm.1 TC0900002727.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 893401 - -
          ABI Mouse Genome Survey Microarray 897911 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline