Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01974644_s1
          See other ZFP691 GE Assays ›
          Gene Symbol:
          Zfp691
          Gene Name:
          zinc finger protein 691
          Gene Aliases:
          6430544H17Rik, Znf691
          Chromosome Location:
          Chr.4: 119169516 - 119174195 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01974644_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp691
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145935.1 NP_001139407.1 2 1641 129
          NM_001145936.1 NP_001139408.1 2 1505 129
          NM_183140.3 NP_898963.2 2 2258 129
          GenBank mRNA AK032431.1 - 2 2257 129
          AK171649.1 - 2 1615 129
          BC108395.1 - 2 1463 30138157 129

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp691

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          biological_process
          nucleic acid binding
          DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23244004 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23244005 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23244006 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23244007 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2080649 AK032431.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2080649 NM_001145935.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2080649 NM_001145936.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2080649 NM_183140.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 785045 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 121232 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 202127 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133413 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2080649 AK032431.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2080649 NM_001145935.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2080649 NM_001145936.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2080649 NM_183140.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23243998 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23243993 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23244005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23244010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23244013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23243996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23244002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23243992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23244000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23244004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23244007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23244011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23244009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23244008 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P133413 - -
          Whole Mouse Genome G4122A A_52_P79923 BC108395.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P79923 AK032431.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P79923 AK171649.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P79923 NM_001145935.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P79923 NM_001145936.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140242 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P133413 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03876326 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03876330 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03876320 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03876321 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03876319 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03876332 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03876334 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23244007 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23244004 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23244006 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23244005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23243992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23243981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23243979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23243970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23243962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23243975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23243984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23243993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23243996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23243986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23243961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23243977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23243972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23243968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23243990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23243983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23243978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23244010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23243967 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03876326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23243970 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03464864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23243972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03603428 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23243989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244002 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03266917 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244012 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23243961 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23243990 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23243998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23243986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23243975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23243965 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23243997 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23243979 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244010 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23243960 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23243996 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23243993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03232337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23243981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23243992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23243962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03876320 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23243982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03364316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23244008 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03876324 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10515846 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515845 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515847 - -
          Affymetrix HT MG-430 PM Array Plate 1456912_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10515844 - -
          Mouse Genome 430 2.0 Array 1456912_at - -
          Mouse Genome 430 2.0 Array 1456912_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 195522_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 195522_at - -
          Affymetrix Mouse Clariom S Array PSR0400027635.mm.1 TC0400003417.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400027635.mm.1 NM_001145935.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027635.mm.1 NM_001145935.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027635.mm.1 NM_001145936.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027635.mm.1 NM_001145936.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027635.mm.1 NM_183140.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027635.mm.1 NM_183140.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027636.mm.1 TC0400003417.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400027636.mm.1 NM_183140.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027636.mm.1 NM_183140.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027637.mm.1 TC0400003417.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400027637.mm.1 NM_001145935.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027637.mm.1 NM_001145935.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027637.mm.1 NM_183140.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027637.mm.1 NM_183140.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027634.mm.1 TC0400003417.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400027634.mm.1 NM_001145935.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027634.mm.1 NM_001145935.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027634.mm.1 NM_001145936.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027634.mm.1 NM_001145936.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027634.mm.1 NM_183140.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027634.mm.1 NM_183140.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027638.mm.1 TC0400003417.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400027638.mm.1 NM_001145936.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027638.mm.1 NM_001145936.1 Refseq
          Mouse Expression Set 430 1456912_PM_at - -
          Mouse Expression Set 430 1456912_at - -
          Affymetrix HT Mouse Genome 430B Array 1456912_at - -
          Affymetrix HT Mouse Genome 430B Array 1456912_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 515044 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3520097 - -
          Illumina mouse-6 v1.0 expression beadchip 3520097 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003520097 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2664559 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2664559 - -
          mouse-6 v1.1 (Illumina) scl23904.2_250-S - -
          Illumina Sentrix mouse-6.v1 bead array 3520097 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl23904.2_250-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3520097 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2813594 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2813594 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline