| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600008901.mm.1 |
NR_040570.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008901.mm.1 |
NR_040570.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008901.mm.1 |
NR_040571.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008901.mm.1 |
NR_040571.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008901.mm.1 |
NR_040572.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008901.mm.1 |
NR_040572.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008901.mm.1 |
TC1600001121.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600008909.mm.1 |
NR_040572.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008909.mm.1 |
NR_040572.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008909.mm.1 |
TC1600001121.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600008902.mm.1 |
NR_040570.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008902.mm.1 |
NR_040570.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008902.mm.1 |
NR_040571.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008902.mm.1 |
NR_040571.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008902.mm.1 |
NR_040572.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008902.mm.1 |
NR_040572.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008902.mm.1 |
TC1600001121.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600008906.mm.1 |
NR_040570.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008906.mm.1 |
NR_040570.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008906.mm.1 |
NR_040571.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008906.mm.1 |
NR_040571.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008906.mm.1 |
NR_040572.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008906.mm.1 |
NR_040572.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008906.mm.1 |
TC1600001121.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600008908.mm.1 |
NR_040570.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008908.mm.1 |
NR_040570.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008908.mm.1 |
NR_040572.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008908.mm.1 |
NR_040572.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008908.mm.1 |
TC1600001121.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600008911.mm.1 |
NR_040570.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008911.mm.1 |
NR_040570.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008911.mm.1 |
NR_040571.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008911.mm.1 |
NR_040571.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600008911.mm.1 |
TC1600001121.mm.1 |
MTA TC |