Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01984314_s1
          See other MRGPRA1 GE Assays ›
          Gene Symbol:
          Mrgpra1
          Gene Name:
          MAS-related GPR, member A1
          Gene Aliases:
          MrgA1
          Chromosome Location:
          Chr.7: 47334875 - 47354240 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01984314_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrgpra1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153095.2 NP_694735.2 2 742 158
          GenBank mRNA AY042191.1 - 2 742 158

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mrgpra1

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          neuropeptide signaling pathway
          chemical synaptic transmission
          signal transducer activity
          G-protein coupled receptor activity
          neuropeptide receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P295996 AY042191.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P295996 BC126873.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P295996 NM_153095.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P295996 AY042191.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P295996 BC126873.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P295996 NM_153095.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 297042 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544892 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 763837 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 696020 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 556606 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 101185 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 36794 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P295996 AY042191.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P295996 BC126873.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P295996 NM_153095.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P295996 AY042191.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P295996 BC126873.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P295996 NM_153095.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P295996 AY042191.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P295996 BC126873.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P295996 NM_153095.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P295996 AY042191.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P295996 BC126873.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P295996 NM_153095.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25103974 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25104008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25103979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25104005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25104007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25103975 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25103997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25103968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25104000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25103991 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25104004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25103978 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01510958 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01510954 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06188087 - -
          Whole Mouse Genome G4122A A_51_P295996 AY042191.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P295996 BC126873.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P295996 NM_153095.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P295996 AY042191.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P295996 BC126873.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P295996 NM_153095.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140891 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01510954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25104005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25104007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25104008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25103939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25104002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25103900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25103955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25103913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25103914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25103998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25103974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25104004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25103899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25103956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25104000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25103882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25103924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25104000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25103975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06666545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25104005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06538881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06666537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25103937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25103939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05908333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25103997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25104004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25103957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25103931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25103979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25103955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25104002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05926556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25103956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06666543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05998232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05908334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25103978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05926557 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10563711 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563710 - -
          Affymetrix HT MG-430 PM Array Plate 1426141_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10563709 - -
          Mouse Genome 430 2.0 Array 1426141_PM_at - -
          Mouse Genome 430 2.0 Array 1426141_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 233221_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 233221_at - -
          Affymetrix Mouse Clariom S Array PSR0700027956.mm.1 NM_153095.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027956.mm.1 NM_153095.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027956.mm.1 TC0700003056.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700027958.mm.1 NM_153095.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027958.mm.1 NM_153095.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027958.mm.1 TC0700003056.mm.1 MTA TC
          Mouse Expression Set 430 1426141_at - -
          Mouse Expression Set 430 1426141_PM_at - -
          Mouse Genome 430A 2.0 Array 1426141_PM_at - -
          Mouse Genome 430A 2.0 Array 1426141_at - -
          Affymetrix HT Mouse Genome 430A Array 1426141_at - -
          Affymetrix HT Mouse Genome 430A Array 1426141_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 831914 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104210373 - -
          Illumina mouse-6 v1.0 expression beadchip 104210373 - -
          Illumina mouse-6 v1.0 expression beadchip 2630465 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002630465 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230705 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229736 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230705 - -
          mouse-6 v1.1 (Illumina) scl31331.2.1_124-S - -
          mouse-6 v1.1 (Illumina) GI_23346520-S - -
          Illumina Sentrix mouse-6.v1 bead array 2630465 - -
          Illumina Sentrix mouse-6.v1 bead array 104210373 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31331.2.1_124-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104210373 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2630465 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1230705 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230705 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0