Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02012751_s1
          See other OLFR476 GE Assays ›
          Gene Symbol:
          Olfr476
          Gene Name:
          olfactory receptor 476
          Gene Aliases:
          MOR204-3
          Chromosome Location:
          Chr.7: 107967399 - 107968331 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02012751_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr476
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146924.1 NP_667135.1 1 324 155

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr476

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          response to stimulus
          detection of chemical stimulus involved in sensory perception of smell
          signal transducer activity
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005611 NM_146924.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005611 AY043306.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112266 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005611 NM_146924.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005611 AY043306.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P344636 NM_146924.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P344636 AY043306.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P112266 - -
          Whole Mouse Genome G4122A A_51_P344636 NM_146924.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P344636 AY043306.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106636 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127994 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P112266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32875659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32876254 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10556122 - -
          Affymetrix Mouse Gene 1.1 ST Array 10556121 - -
          Affymetrix Mouse Clariom S Array PSR0700013263.mm.1 TC0700001588.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700013263.mm.1 NM_146924.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700013263.mm.1 NM_146924.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 674832 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2993799 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2993799 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline