Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02013620_g1
          See other MORF4L2 GE Assays ›
          Gene Symbol:
          Morf4l2
          Gene Name:
          mortality factor 4 like 2
          Gene Aliases:
          2410017O14Rik, Mrgx, Sid393p, mKIAA0026
          Chromosome Location:
          Chr.X: 136732942 - 136743686 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02013620_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Morf4l2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001168225.1 NP_001161697.1 4 1532 75
          NM_001168226.1 NP_001161698.1 4 1465 75
          NM_001168227.1 NP_001161699.1 4 1417 75
          NM_001168228.1 NP_001161700.1 4 1416 75
          NM_001168229.1 NP_001161701.1 3 1388 75
          NM_001168230.1 NP_001161702.1 3 1326 75
          NM_019768.4 NP_062742.4 4 1539 75
          GenBank mRNA AB025049.1 - 2 1227 75
          AK075946.1 - 4 1392 75
          AK088849.1 - 1 1246 75
          AK089963.1 - 4 1393 75
          AK129038.1 - 4 1373 75
          AK131944.1 - 4 1392 75
          AK145686.1 - 4 1392 75
          AK150401.1 - 4 1392 75
          AK150670.1 - 4 1392 75
          AK151570.1 - 4 1392 75
          AK153146.1 - 4 1388 75
          AK159145.1 - 4 1441 75
          AK159179.1 - 4 1391 75
          AK160012.1 - 4 1392 75
          AK161610.1 - 4 1418 75
          AK164433.1 - 4 1392 75
          AK165985.1 - 4 1392 75
          AK166637.1 - 4 1418 75
          AK211563.1 - 1 252 75
          AK211831.1 - 1 352 75
          BC075653.1 - 4 1539 30539330 75
          BC088731.1 - 1 1173 5340043 75

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Morf4l2

          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s)

          DNA repair
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          cellular response to DNA damage stimulus
          chromatin modification
          regulation of growth

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32682198 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1969876 NM_001168225.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1969882 BC075653.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1969882 BC088731.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1969882 AK164433.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1969882 AK165985.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1969882 AK166637.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 920571 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 149528 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 48522 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 898556 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 675035 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 5934 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125934 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1969876 NM_001168225.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1969882 BC075653.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1969882 BC088731.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1969882 AK164433.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1969882 AK165985.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1969882 AK166637.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416387 BC075653.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416387 BC088731.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416387 AK164433.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416387 AK165985.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416387 AK166637.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32682192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32682180 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32682187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32682196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32682185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32682190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32682198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32682186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32682189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32682193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32682182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32682197 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125934 - -
          Whole Mouse Genome G4122A A_51_P416387 BC075653.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416387 BC088731.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416387 AK164433.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416387 AK165985.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416387 AK166637.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136074 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125934 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14574248 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32682198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32682159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682143 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682151 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14956663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14626938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14793552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682122 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682079 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14793563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32682160 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10606879 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606877 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606878 - -
          Affymetrix HT MG-430 PM Array Plate 1415778_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1439432_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1456743_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1439413_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10606876 - -
          Affymetrix Mouse Gene 1.1 ST Array 10601880 - -
          Mouse Genome 430 2.0 Array 1439432_PM_x_at - -
          Mouse Genome 430 2.0 Array 1456743_x_at - -
          Mouse Genome 430 2.0 Array 1415778_at - -
          Mouse Genome 430 2.0 Array 1439413_x_at - -
          Mouse Genome 430 2.0 Array 1415778_PM_at - -
          Mouse Genome 430 2.0 Array 1456743_PM_x_at - -
          Mouse Genome 430 2.0 Array 1439413_PM_x_at - -
          Mouse Genome 430 2.0 Array 1439432_x_at - -
          Mouse Expression Set 430 1439432_PM_x_at - -
          Mouse Expression Set 430 1439432_x_at - -
          Mouse Expression Set 430 1415778_PM_at - -
          Mouse Expression Set 430 1456743_PM_x_at - -
          Mouse Expression Set 430 1415778_at - -
          Mouse Expression Set 430 1439413_x_at - -
          Mouse Expression Set 430 1456743_x_at - -
          Mouse Expression Set 430 1439413_PM_x_at - -
          Murine Genome U74v2 Set 96633_s_at - -
          Murine Genome U74v2 Set 96632_at - -
          Murine Genome U74v2 Set 96633_PM_s_at - -
          Murine Genome U74v2 Set 96632_PM_at - -
          Murine Genome U74v2 Set 169402_PM_r_at - -
          Murine Genome U74v2 Set 169402_r_at - -
          Mouse Genome 430A 2.0 Array 1415778_at - -
          Mouse Genome 430A 2.0 Array 1439432_x_at - -
          Mouse Genome 430A 2.0 Array 1456743_x_at - -
          Mouse Genome 430A 2.0 Array 1439413_x_at - -
          Mouse Genome 430A 2.0 Array 1439432_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1439413_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1415778_PM_at - -
          Mouse Genome 430A 2.0 Array 1456743_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1415778_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1439413_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1439413_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1415778_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 897671 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6450133 - -
          Illumina mouse-6 v1.0 expression beadchip 6450133 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006450133 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1238425 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1238425 - -
          mouse-6 v1.1 (Illumina) scl056397.1_158-S - -
          Illumina Sentrix mouse-6.v1 bead array 6450133 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl056397.1_158-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6450133 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2599618 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2599618 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl056397.1_158-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline