Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02019269_g1
          See other ODC1 GE Assays ›
          Gene Symbol:
          Odc1
          Gene Name:
          ornithine decarboxylase, structural 1
          Gene Aliases:
          ODC
          Chromosome Location:
          Chr.12: 17544873 - 17551502 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02019269_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Odc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013614.2 NP_038642.2 6 947 115
          GenBank mRNA AK077679.1 - 6 849 115
          AK139610.1 - 6 849 115
          AK150283.1 - 6 851 115
          AK150343.1 - 6 850 115
          AK152525.1 - 6 849 115
          AK166093.1 - 6 854 115
          AK168219.1 - 6 850 115
          AK172568.1 - 6 849 115
          AK209360.1 - 2 214 115
          BC033264.1 - 6 838 2645291 115
          BC059826.1 - 6 828 6808137 115
          BC083122.1 - 6 835 6329274 115
          EU684749.1 - 4 537 115
          M20617.2 - 4 553 115
          S64539.1 - 6 847 115

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Odc1

          Panther Classification:

          Molecular Function -

          decarboxylase

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          polyamine metabolic process
          polyamine biosynthetic process
          positive regulation of cell proliferation
          putrescine biosynthetic process
          putrescine biosynthetic process from ornithine
          regulation of protein catabolic process
          catalytic activity
          ornithine decarboxylase activity
          lyase activity
          carboxy-lyase activity
          protein homodimerization activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200001238.mm.1 NM_013614.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001238.mm.1 NM_013614.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001238.mm.1 TC1200000142.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200001246.mm.1 NM_013614.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001246.mm.1 NM_013614.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001246.mm.1 TC1200000142.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200001239.mm.1 NM_013614.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001239.mm.1 NM_013614.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001239.mm.1 TC1200000142.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200001244.mm.1 NM_013614.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001244.mm.1 NM_013614.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001244.mm.1 TC1200000142.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200001248.mm.1 NM_013614.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001248.mm.1 NM_013614.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001248.mm.1 TC1200000142.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200001235.mm.1 NM_013614.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001235.mm.1 NM_013614.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001235.mm.1 TC1200000142.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200001241.mm.1 NM_013614.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001241.mm.1 NM_013614.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001241.mm.1 TC1200000142.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200001245.mm.1 NM_013614.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001245.mm.1 NM_013614.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001245.mm.1 TC1200000142.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200001243.mm.1 NM_013614.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001243.mm.1 NM_013614.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001243.mm.1 TC1200000142.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200001240.mm.1 NM_013614.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001240.mm.1 NM_013614.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001240.mm.1 TC1200000142.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200001242.mm.1 NM_013614.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001242.mm.1 NM_013614.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001242.mm.1 TC1200000142.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200001247.mm.1 NM_013614.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001247.mm.1 NM_013614.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001247.mm.1 TC1200000142.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 899662 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline