Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02021221_s1
          See other GM7968 GE Assays ›
          Gene Symbol:
          Gm7968
          Gene Name:
          predicted gene 7968
          Gene Aliases:
          EG666183
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02021221_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm7968
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK036791.1 - 1 2161 100

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2180521 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P272735 AK036791.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P272735 AK036791.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P272735 AK036791.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P272735 AK036791.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P30134009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30134006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30134001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30134043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30134040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12036534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30134007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30134010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30134023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12200666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11933968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30134028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30134041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30134004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12131077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12055395 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30134026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12430691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30134016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30134032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30134011 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12330287 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30134036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30134038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12014787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12357675 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 713112 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline