Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02022226_s1
          See other DEFB41 GE Assays ›
          Gene Symbol:
          Defb41
          Gene Name:
          defensin beta 41
          Gene Aliases:
          9230102D03Rik, Bd-41, Defb16, Gm15386, OTTMUSG00000020946, OTTMUSG00000020948
          Chromosome Location:
          Chr.1: 18250978 - 18265138 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02022226_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Defb41
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_183124.3 NP_898947.1 2 391 139
          GenBank mRNA AK020304.1 - 2 392 139
          DQ012027.1 - 2 391 139

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Defb41

          Panther Classification:

          Molecular Function -

          antimicrobial response protein

          Gene Ontology Categories:

          Function(s) Process(es)

          defense response
          biological_process
          defense response to bacterium
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P307828 DQ012027.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P307828 NM_183124.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P307828 AK020304.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P307828 DQ012027.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P307828 NM_183124.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P307828 AK020304.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 145667 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 55327 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 118914 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 123706 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 382618 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 419720 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 850182 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 98052 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 160006 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 111908 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142548 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154104 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P307828 DQ012027.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P307828 NM_183124.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P307828 AK020304.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P307828 DQ012027.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P307828 NM_183124.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P307828 AK020304.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20062626 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20062624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20062630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20062627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20062621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20062616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20062620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20062619 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00018743 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154104 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142548 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04027008 - -
          Whole Mouse Genome G4122A A_51_P307828 DQ012027.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P307828 NM_183124.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P307828 AK020304.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P307828 DQ012027.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P307828 NM_183124.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P307828 AK020304.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132261 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154104 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142548 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00085465 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00085461 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00410762 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00085474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20062646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20062570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20062639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00722244 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20062569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20062626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00649852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20062624 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20062577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20062641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20062635 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20062562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20062620 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20062561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20062619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20062575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20062571 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20062628 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20062637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20062565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00389745 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20062568 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20062616 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20062621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20062642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20062574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20062622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20062640 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00187907 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20062578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20062627 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20062567 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10353339 - -
          Affymetrix Mouse Gene 1.0 ST Array 10353337 - -
          Affymetrix Mouse Gene 1.0 ST Array 10353340 - -
          Affymetrix HT MG-430 PM Array Plate 1440767_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10353336 - -
          Mouse Genome 430 2.0 Array 1440767_at - -
          Mouse Genome 430 2.0 Array 1440767_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100016507.mm.1 NM_183124.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100016507.mm.1 NM_183124.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100016507.mm.1 TC0100002022.mm.1 MTA TC
          Mouse Expression Set 430 1440767_PM_at - -
          Mouse Expression Set 430 1440767_at - -
          Murine Genome U74v2 Set 168423_f_at - -
          Murine Genome U74v2 Set 168423_PM_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1440767_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440767_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5080113 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005080113 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1229247 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229247 - -
          mouse-6 v1.1 (Illumina) scl16958.3.1_50-S - -
          Illumina Sentrix mouse-6.v1 bead array 5080113 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl16958.3.1_50-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5080113 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1229247 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1229247 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline