Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02022307_s1
          See other A330069K06RIK GE Assays ›
          Gene Symbol:
          A330069K06Rik
          Gene Name:
          RIKEN cDNA A330069K06 gene
          Gene Aliases:
          Gm618
          Chromosome Location:
          Chr.3: 80204030 - 80212442 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02022307_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A330069K06Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK039597.1 - 660 95

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2230663 AK039597.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 329409 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 83796 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 16702 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 785104 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 481350 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 604614 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 721681 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 457412 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 518841 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 614851 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 851080 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 292461 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 274375 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 147343 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 418174 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 41503 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 387993 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 795422 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 15781 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 303758 AF220205.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 303758 AK005453.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 303758 AK011086.2 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 303758 BC025098.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 303758 NM_022999.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 575737 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 743583 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 310941 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 616806 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 816433 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 619530 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 636179 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 454408 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 137932 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753232 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 195822 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 723183 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 943101 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 590844 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 763488 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 756321 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 264270 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 942773 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 127885 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 946019 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 71111 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 435494 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 334278 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 157721 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 884639 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 950768 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 342768 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 733736 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 164827 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 193156 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 852716 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 890586 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 140599 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 698990 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 796788 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 98464 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 59505 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 2634 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 117498 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 135010 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 21837 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 89565 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 161927 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00680782 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04961533 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04961652 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04961402 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04961434 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04961605 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04961471 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04961570 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04961516 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04961675 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04961703 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04961561 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04961471 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04961652 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04961703 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04961605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22267404 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22267402 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22267403 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22267399 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22267405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22267397 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 606470 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline