Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02023034_s1
          See other RNASE9 GE Assays ›
          Gene Symbol:
          Rnase9
          Gene Name:
          ribonuclease, RNase A family, 9 (non-active)
          Gene Aliases:
          ESRL, Rak1
          Chromosome Location:
          Chr.14: 51038459 - 51041905 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02023034_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnase9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_183032.2 NP_898853.2 2 1033 86
          GenBank mRNA AK078986.1 - 2 1034 86
          AY226989.1 - 2 1034 86
          AY303816.1 - 2 1033 86

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rnase9

          Gene Ontology Categories:

          Function(s) Process(es)

          RNA phosphodiester bond hydrolysis
          molecular_function
          nucleic acid binding
          endonuclease activity
          ribonuclease activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P345372 BC107218.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P345372 NM_183032.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P345372 AK078986.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P345372 AY226989.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P345372 AY303816.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P345372 BC107218.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P345372 NM_183032.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P345372 AK078986.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P345372 AY226989.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P345372 AY303816.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 671142 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 41151 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 852932 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 94704 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P130685 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P345372 BC107218.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P345372 NM_183032.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P345372 AK078986.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P345372 AY226989.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P345372 AY303816.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P345372 BC107218.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P345372 NM_183032.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P345372 AK078986.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P345372 AY226989.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P345372 AY303816.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599729 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599723 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599712 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599725 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599717 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599709 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599708 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599715 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29599727 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02849437 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P130685 - -
          Whole Mouse Genome G4122A A_52_P345372 BC107218.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P345372 NM_183032.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P345372 AK078986.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P345372 AY226989.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P345372 AY303816.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P345372 BC107218.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P345372 NM_183032.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P345372 AK078986.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P345372 AY226989.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P345372 AY303816.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134965 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102899 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P130685 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11277826 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11277818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29599744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599693 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599669 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11277808 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11386455 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599706 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599712 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599745 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599730 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599707 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11277798 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599735 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599686 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11347939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599731 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599761 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599689 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11519015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11277819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11386461 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599766 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11386453 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599726 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599654 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11302715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11578023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599656 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11756335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599756 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599747 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599684 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599674 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599759 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11756329 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11756334 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599742 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11485970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599692 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599732 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599714 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599738 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29599736 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10419556 - -
          Affymetrix Mouse Gene 1.0 ST Array 10419555 - -
          Affymetrix Mouse Gene 1.0 ST Array 10419554 - -
          Affymetrix Mouse Gene 1.1 ST Array 10419553 - -
          Affymetrix Mouse Clariom S Array PSR1400015019.mm.1 NM_183032.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015019.mm.1 NM_183032.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015019.mm.1 TC1400001995.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400015018.mm.1 NM_183032.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015018.mm.1 NM_183032.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015018.mm.1 TC1400001995.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400015022.mm.1 NM_183032.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015022.mm.1 NM_183032.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015022.mm.1 TC1400001995.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 645506 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4570286 - -
          Illumina mouse-6 v1.0 expression beadchip 4570286 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004570286 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1219190 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1219190 - -
          mouse-6 v1.1 (Illumina) scl45608.2.1_315-S - -
          Illumina Sentrix mouse-6.v1 bead array 4570286 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl45608.2.1_315-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4570286 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1219190 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1219190 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0