Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02024151_s1
          See other ZFP580 GE Assays ›
          Gene Symbol:
          Zfp580
          Gene Name:
          zinc finger protein 580
          Gene Aliases:
          1500015O20Rik, AI838225, Znf580
          Chromosome Location:
          Chr.7: 5051532 - 5053723 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02024151_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp580
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026900.1 NP_081176.1 2 1040 107
          GenBank mRNA AK005257.1 - 2 1035 107
          BC066001.1 - 2 988 5700640 107

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of endothelial cell proliferation
          positive regulation of leukocyte chemotaxis
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          chemotaxis
          inflammatory response
          positive regulation of endothelial cell migration
          positive regulation of gene expression
          positive regulation of interleukin-8 production
          positive regulation of cellular component movement
          cellular response to hydrogen peroxide
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          nucleic acid binding
          DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353882 NM_026900.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353882 AK005257.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353882 BC066001.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353882 NM_026900.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353882 AK005257.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353882 BC066001.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 274517 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 578741 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156098 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353882 NM_026900.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353882 AK005257.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353882 BC066001.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353882 NM_026900.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353882 AK005257.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353882 BC066001.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353882 NM_026900.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353882 AK005257.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353882 BC066001.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353882 NM_026900.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353882 AK005257.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353882 BC066001.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116836 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10549761 - -
          Affymetrix HT MG-430 PM Array Plate 1429634_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10549760 - -
          Mouse Genome 430 2.0 Array 1429634_PM_at - -
          Mouse Genome 430 2.0 Array 1429634_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68992_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68992_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700000632.mm.1 TC0700000055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700000632.mm.1 NM_026900.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000632.mm.1 NM_026900.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000634.mm.1 TC0700000055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700000634.mm.1 NM_026900.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000634.mm.1 NM_026900.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000633.mm.1 TC0700000055.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700000633.mm.1 NM_026900.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000633.mm.1 NM_026900.1 Refseq
          Mouse Expression Set 430 1429634_at - -
          Mouse Expression Set 430 1429634_PM_at - -
          Murine Genome U74v2 Set 171565_PM_i_at - -
          Murine Genome U74v2 Set 171565_i_at - -
          Murine Genome U74v2 Set 169005_f_at - -
          Murine Genome U74v2 Set 169005_PM_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1429634_at - -
          Affymetrix HT Mouse Genome 430B Array 1429634_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 885152 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 101780113 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2446373 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230154 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2446373 - -
          mouse-6 v1.1 (Illumina) scl00066.1_84-S - -
          Illumina Sentrix mouse-6.v1 bead array 101780113 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101780113 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2433470 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0