Search
Search

Tcstv3| Gene Symbol: Tcstv3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153523.3 | NP_705743.1 | 2 | 721 | 125 | |
| GenBank mRNA | AF067060.1 | - | 719 | 125 | ||
| BC125501.1 | - | 2 | 704 | 40130016 | 125 | |
| BC125503.1 | - | 2 | 704 | 40130018 | 125 | |
Set Membership: |
|